Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CPA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020361.4 | NP_065094.3 | 7, 8 | 1011 | ||
XM_011517570.2 | XP_011515872.1 | 6, 7 | 1425 | |||
XM_017013646.1 | XP_016869135.1 | 7, 8 | 653 | |||
GenBank mRNA | AF221594.1 | - | 7, 8 | 968 | ||
AY044833.1 | - | 6, 7 | 844 | |||
BC033684.1 | - | 7, 8 | 971 | 5193403 |