Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PTPN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014369.3 | NP_055184.2 | 2 | 294 | ||
XM_006712417.1 | XP_006712480.1 | 2 | 289 | |||
XM_017003798.1 | XP_016859287.1 | 2 | 245 | |||
GenBank mRNA | DN995571.1 | - | 2 | 302 | ||
X79568.1 | - | 2 | 236 |