Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288607.1 | NP_001275536.1 | 6 | 1368 | ||
NM_001288608.1 | NP_001275537.1 | 6 | 1368 | |||
NM_019032.5 | NP_061905.2 | 6 | 1368 | |||
NM_025008.4 | NP_079284.2 | 6 | 1368 | |||
XM_011509644.2 | XP_011507946.1 | 5 | 1330 | |||
XM_011509645.2 | XP_011507947.1 | 5 | 1327 | |||
XM_011509648.2 | XP_011507950.1 | 5 | 1284 | |||
XM_011509649.2 | XP_011507951.1 | 5 | 1338 | |||
XM_011509650.2 | XP_011507952.1 | 5 | 1338 | |||
XM_017001506.1 | XP_016856995.1 | 6 | 1739 | |||
GenBank mRNA | AY358122.1 | - | 5 | 1182 | ||
BC140800.1 | - | 5 | 1220 | 9021668 | ||
BC144581.1 | - | 5 | 1220 | 9053111 | ||
BX370116.2 | - | 1 | 633 |