Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IL17RD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318864.1 | NP_001305793.1 | 8 | 844 | ||
NM_017563.4 | NP_060033.3 | 7 | 835 | |||
XM_005265238.4 | XP_005265295.1 | 7 | 1046 | |||
XM_006713209.3 | XP_006713272.1 | 9 | 973 | |||
XM_011533849.2 | XP_011532151.2 | 7 | 630 | |||
GenBank mRNA | AF458067.1 | - | 7 | 722 | ||
AF494208.1 | - | 7 | 811 | |||
AF494211.1 | - | 8 | 812 | |||
AK093074.1 | - | 8 | 811 | |||
AK302045.1 | - | 7 | 661 | |||
AL133097.1 | - | 3 | 197 | |||
AL833913.1 | - | 7 | 635 | |||
AY358774.1 | - | 7 | 758 | |||
AY489047.1 | - | 8 | 837 | |||
BC038369.1 | - | 8 | 833 | |||
BC111702.1 | - | 7 | 662 | 40038598 |