Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KANSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017822.3 | NP_060292.3 | 7, 8 | 1048 | ||
GenBank mRNA | AK022732.1 | - | 6, 7 | 786 | ||
AK094528.1 | - | 6, 7 | 1571 | |||
AL834467.1 | - | 4, 5 | 467 | |||
BC009746.2 | - | 7, 8 | 1016 | 3503970 | ||
BC013900.2 | - | 6, 7 | 968 | 3928141 | ||
BG760446.1 | - | 2, 3 | 218 |