Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286212.1 | NP_001273141.1 | 6 | 1149 | ||
NM_001286213.1 | NP_001273142.1 | 6 | 1116 | |||
NM_032256.2 | NP_115632.1 | 7 | 1249 | |||
XM_011538831.2 | XP_011537133.1 | 7 | 1258 | |||
XM_011538832.2 | XP_011537134.1 | 7 | 933 | |||
XM_011538833.2 | XP_011537135.1 | 6 | 750 | |||
XM_017020022.1 | XP_016875511.1 | 7 | 840 | |||
XM_017020023.1 | XP_016875512.1 | 8 | 1081 | |||
GenBank mRNA | AK057287.1 | - | 7 | 968 | ||
AK097511.1 | - | 2 | 194 | |||
AL136855.1 | - | 7 | 951 | |||
BC030284.1 | - | 6 | 857 | 5240522 | ||
BC060798.1 | - | 7 | 1012 | 30348706 | ||
DA007410.1 | - | 6 | 557 |