Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DGAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253891.1 | NP_001240820.1 | 3 | 540 | ||
NM_032564.4 | NP_115953.2 | 4 | 669 | |||
XM_011545304.2 | XP_011543606.1 | 4 | 373 | |||
GenBank mRNA | AB048286.2 | - | 4 | 640 | ||
AB073384.1 | - | 3 | 217 | |||
AF384161.2 | - | 4 | 417 | |||
AL834287.1 | - | 4 | 548 | |||
AY358532.1 | - | 4 | 594 | |||
AY780647.1 | - | 3 | 272 | |||
BC015234.1 | - | 4 | 658 | 3903313 |