Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCDC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144719.3 | NP_653320.3 | 10, 11 | 1460 | ||
XM_005264898.3 | XP_005264955.1 | 10, 11 | 1460 | |||
XM_011533418.2 | XP_011531720.1 | 10, 11 | 1460 | |||
XM_011533419.2 | XP_011531721.1 | 6, 7 | 1471 | |||
XM_017005781.1 | XP_016861270.1 | 6, 7 | 858 | |||
GenBank mRNA | BC036050.1 | - | 10, 11 | 1462 | 5270468 |