Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320435.1 | NP_001307364.1 | 2 | 256 | ||
NM_001320436.1 | NP_001307365.1 | 2 | 256 | |||
NM_198154.2 | NP_937797.1 | 2 | 256 | |||
XM_017024565.1 | XP_016880054.1 | 2 | 278 | |||
XM_017024566.1 | XP_016880055.1 | 2 | 278 | |||
XM_017024567.1 | XP_016880056.1 | 2 | 278 | |||
XM_017024568.1 | XP_016880057.1 | 2 | 278 | |||
XM_017024569.1 | XP_016880058.1 | 2 | 278 | |||
XM_017024571.1 | XP_016880060.1 | 2 | 424 | |||
GenBank mRNA | AY358218.1 | - | 2 | 214 | ||
BC040900.1 | - | 2 | 256 | |||
BC107110.2 | - | 2 | 214 | 40022149 |