Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FOLH1 SIRNA Assays ›
Gene Symbol:
FOLH1
Gene Name:
folate hydrolase (prostate-specific membrane antigen) 1
Gene Aliases:
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
Chromosome Location:
Chr.11: 49145094 - 49208706 on Build GRCh38
UniGene ID:
Hs.645352
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s5329
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FOLH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001014986.1
NP_001014986.1
3
589
NM_001193471.1
NP_001180400.1
4
686
NM_001193472.1
NP_001180401.1
4
686
NM_004476.1
NP_004467.1
3
589
XM_011519958.2
XP_011518260.2
4
723
XM_017017432.1
XP_016872921.1
4
721
XM_017017433.1
XP_016872922.1
4
721
XM_017017434.1
XP_016872923.1
4
387
XM_017017435.1
XP_016872924.1
4
386
XM_017017436.1
XP_016872925.1
3
621
XM_017017437.1
XP_016872926.1
3
348
XM_017017438.1
XP_016872927.1
3
621
XM_017017439.1
XP_016872928.1
4
422
XM_017017440.1
XP_016872929.1
3
292
XM_017017441.1
XP_016872930.1
3
233
XM_017017442.1
XP_016872931.1
3
359
XM_017017443.1
XP_016872932.1
3
243
XM_017017444.1
XP_016872933.1
3
523
XM_017017445.1
XP_016872934.1
4
650
XM_017017446.1
XP_016872935.1
3
621
XM_017017447.1
XP_016872936.1
3
291
XM_017017448.1
XP_016872937.1
3
283
XM_017017449.1
XP_016872938.1
3
523
XM_017017450.1
XP_016872939.1
4
650
XM_017017451.1
XP_016872940.1
3
292
GenBank mRNA
AF176574.1
-
3
466
AJ007318.1
-
4
343
AK295470.1
-
4
618
AK300532.1
-
3
521
AK312366.1
-
3
517
AY101595.1
-
3
328
BC025672.1
-
3
487
5202715
DQ088979.1
-
3
451
EF488811.2
-
4
388
EF488812.1
-
4
430
M99487.1
-
3
589
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
folic acid-containing compound metabolic process
cellular amino acid biosynthetic process
C-terminal protein deglutamylation
metallocarboxypeptidase activity
peptidase activity
dipeptidase activity
metal ion binding
Ac-Asp-Glu binding
tetrahydrofolyl-poly(glutamate) polymer binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0