Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KGFLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003674.2 | 1 | 186 | |||
NR_047527.1 | 1 | 186 | ||||
GenBank mRNA | AF523265.1 | - | 1 | 281 | ||
AY098593.1 | - | 1 | 177 | |||
BC101842.1 | - | 1 | 186 | |||
BC112977.1 | - | 1 | 184 | |||
BC112978.1 | - | 1 | 184 |
Gene Symbol: KGFLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003670.1 | 1 | 285 |
Gene Symbol: LOC102724238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK290223.1 | - | 1 | 193 |