Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ctnnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085448.1 | NP_001078917.1 | 6 | 1749 | ||
NM_001085449.1 | NP_001078918.1 | 5 | 1460 | |||
NM_001085450.1 | NP_001078919.1 | 6 | 1749 | |||
NM_001085453.1 | NP_001078922.1 | 6 | 1519 | |||
NM_007615.4 | NP_031641.2 | 7 | 1867 | |||
GenBank mRNA | AB093237.1 | - | 7 | 1583 | ||
AK133193.1 | - | 6 | 1470 | |||
AK148871.1 | - | 2 | 530 | |||
AK161790.1 | - | 7 | 1643 | |||
AK164485.1 | - | 6 | 1734 | |||
AK168453.1 | - | 5 | 1458 | |||
AK206736.1 | - | 1 | 94 | |||
BC043108.1 | - | 6 | 1684 | 6406902 | ||
BC046589.1 | - | 5 | 1260 | |||
BC054544.1 | - | 7 | 1800 | 6408956 | ||
Z17804.1 | - | 1662 |