Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Epas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010137.3 | NP_034267.3 | 12 | 2346 | ||
GenBank mRNA | AF045160.1 | - | 12 | 2110 | ||
AK136911.1 | - | 12 | 2350 | |||
AK141185.1 | - | 12 | 2081 | |||
AK158430.1 | - | 12 | 2510 | |||
BC057870.1 | - | 12 | 2068 | 5032291 | ||
D89787.1 | - | 12 | 2308 | |||
U81983.1 | - | 12 | 2029 |