Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sept9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113486.1 | NP_001106958.1 | 4 | 982 | ||
NM_001113487.1 | NP_001106959.1 | 3 | 1162 | |||
NM_001113488.1 | NP_001106960.1 | 2 | 331 | |||
NM_017380.2 | NP_059076.1 | 2 | 312 | |||
GenBank mRNA | AJ250723.1 | - | 2 | 318 | ||
AK031757.1 | - | 2 | 207 | |||
AK122415.2 | - | 3 | 956 | |||
AK141312.1 | - | 4 | 923 | |||
AK164581.1 | - | 2 | 217 | |||
BC046524.1 | - | 4 | 982 | 5290621 | ||
BC094455.1 | - | 5 | 686 | |||
BU612221.1 | - | 2 | 314 | |||
BY772945.1 | - | 2 | 299 |