Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ivd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019826.3 | NP_062800.1 | 6 | 629 | ||
GenBank mRNA | AF225989.1 | - | 6 | 608 | ||
AK005024.1 | - | 6 | 648 | |||
AK017660.1 | - | 6 | 615 | |||
AK077416.1 | - | 6 | 636 | |||
BC018325.1 | - | 6 | 619 | 4192286 | ||
BC027198.1 | - | 6 | 629 | 4237982 |