Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adamdec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021475.2 | NP_067450.1 | 5, 6 | 467 | ||
GenBank mRNA | AJ242912.1 | - | 5, 6 | 467 | ||
AK156586.1 | - | 5, 6 | 482 | |||
BC026542.1 | - | 5, 6 | 490 | |||
BC046324.1 | - | 5, 6 | 458 | 6394673 |