Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 2810428I15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025577.2 | NP_079853.1 | 2 | 128 | ||
GenBank mRNA | AK013181.1 | - | 2 | 129 | ||
AK160511.1 | - | 2 | 72 | |||
BC028495.1 | - | 2 | 134 | 3370477 |