Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rab39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175122.6 | NP_780331.1 | 2 | 533 | ||
GenBank mRNA | AB232636.1 | - | 2 | 309 | ||
AK018257.1 | - | 2 | 517 | |||
AK039013.1 | - | 2 | 507 | |||
AK048981.1 | - | 2 | 507 | |||
AK160737.1 | - | 2 | 507 | |||
BC050853.1 | - | 2 | 527 | 6517891 | ||
BC052472.1 | - | 2 | 458 | 6833461 |