Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mfsd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028140.4 | NP_082416.2 | 12 | 1629 | ||
GenBank mRNA | AK013144.1 | - | 12 | 1521 | ||
AK032080.1 | - | 12 | 1650 | |||
AK035291.1 | - | 12 | 1593 | |||
AK077957.1 | - | 12 | 1630 | |||
BC010483.1 | - | 12 | 1478 | |||
BC113183.1 | - | 12 | 1560 | 40061891 | ||
BC113790.1 | - | 12 | 1560 | 40061889 |