- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Kremen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028416.2 | NP_082692.1 | 2, 3 | 262 | ||
GenBank mRNA | AJ457192.1 | - | 2, 3 | 274 |