Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MYO1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303279.1 | NP_001290208.1 | 2 | 415 | ||
NM_001303280.1 | NP_001290209.1 | 2 | 415 | |||
NM_015194.2 | NP_056009.1 | 2 | 415 | |||
XM_017024685.1 | XP_016880174.1 | 2 | 386 | |||
GenBank mRNA | AB018270.2 | - | 2 | 126 | ||
AK127942.1 | - | 2 | 386 | |||
BC146763.1 | - | 2 | 126 | 9007203 |