Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rab3il1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144538.2 | NP_653121.1 | 4 | 497 | ||
GenBank mRNA | AK004767.1 | - | 4 | 492 | ||
AK132764.1 | - | 7 | 1204 | |||
AK161298.1 | - | 7 | 1204 | |||
BC020147.1 | - | 4 | 462 | 3708973 |