Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bcl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029850.3 | NP_084126.1 | 2 | 312 | ||
GenBank mRNA | AK014498.1 | - | 2 | 255 | ||
AK044782.1 | - | 2 | 283 | |||
AK086803.1 | - | 2 | 309 | |||
AK157720.1 | - | 2 | 261 | |||
BC017640.1 | - | 2 | 226 | 4482250 | ||
BC038481.1 | - | 2 | 237 | 4501205 | ||
BC046777.1 | - | 2 | 142 | 5709511 | ||
BY271432.1 | - | 2 | 312 |