Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mtus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005863.2 | NP_001005863.1 | 13, 14 | 3826 | ||
NM_001005864.3 | NP_001005864.1 | 11, 12 | 1432 | |||
NM_001005865.3 | NP_001005865.2 | 9, 10 | 1445 | |||
NM_001286413.1 | NP_001273342.1 | 13, 14 | 3708 | |||
GenBank mRNA | AF173380.1 | - | 9, 10 | 1284 | ||
AF493235.1 | - | 9, 10 | 1349 | |||
AK030510.1 | - | 9, 10 | 1409 | |||
AK031693.1 | - | 9, 10 | 1379 | |||
AK035576.1 | - | 7, 8 | 813 | |||
AK129324.1 | - | 7, 8 | 3922 | |||
AY246699.1 | - | 9, 10 | 1107 | |||
AY626781.1 | - | 9, 10 | 1442 | |||
AY626782.1 | - | 11, 12 | 1430 | |||
AY626783.1 | - | 12, 13 | 3648 | |||
BC030860.1 | - | 1, 2 | 99 | |||
BC041777.1 | - | 12, 13 | 2491 | 4240274 | ||
BC042206.1 | - | 12, 13 | 2491 | 4240274 | ||
BC043321.1 | - | 11, 12 | 1396 | |||
BC089009.1 | - | 13, 14 | 3708 | 30543240 |