Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc38a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166456.1 | NP_001159928.1 | 10, 11 | 1128 | ||
NM_001166458.1 | NP_001159930.1 | 10, 11 | 1335 | |||
NM_134086.4 | NP_598847.2 | 10, 11 | 1821 | |||
GenBank mRNA | AF184240.2 | - | 9, 10 | 913 | ||
AK029189.1 | - | 10, 11 | 998 | |||
AK034130.1 | - | 10, 11 | 1823 | |||
AK035098.1 | - | 9, 10 | 840 | |||
AK047459.1 | - | 10, 11 | 1352 | |||
AK049173.1 | - | 10, 11 | 1036 | |||
AK049294.1 | - | 10, 11 | 1101 | |||
AK050914.1 | - | 10, 11 | 1036 | |||
AK081724.1 | - | 10, 11 | 1337 | |||
AK082683.1 | - | 10, 11 | 1038 | |||
AK162612.1 | - | 10, 11 | 1186 | |||
AK163914.1 | - | 5, 6 | 439 | |||
AK165188.1 | - | 10, 11 | 1388 | |||
AK217621.1 | - | 3, 4 | 199 | |||
BC030378.1 | - | 10, 11 | 1037 | 5375319 |