Type:
Whole Allergen
Whole Allergen
Whole Allergen
Yeast
Inhalation
Saccharomycetaceae
Saccharomyces cerevisiae
Saccharomyces cerevisiae
Baker's yeast, Brewer’s yeast
Saccharomyces cerevisiae is a unicellular fungus known as baker's yeast or brewer's yeast. It is an edible budding yeast that is considered safe and therefore possesses various applications in food, beverage, biofuels, and pharmaceutical industry. It occurs naturally in plants and soil. S. cerevisiae is round-to-oval in shape and reproduces through a budding mechanism. Different strains of S. cerevisiae are found in various parts of the world. Allergic reactions due to baker's yeast are rarely reported. S. cerevisiae contains both inhalant and food allergens. It may induce allergic respiratory symptoms, most predominantly baker’s asthma, and allergic rhinitis and also hypersensitivity pneumonitis on inhalation. Moreover, atopic dermatitis and anaphylaxis on ingestion of yeast have been rarely reported in baker’s yeast sensitized individuals. Additionally, anti- S. cerevisiae antibodies (ASCA) are considered as biomarkers for patients suffering from irritable bowel disease. The major allergen identified in S. cerevisiae is Sac c enolase, which is a cross-reactive allergen. Cross-reactivity has been observed between other yeast and mold species. S. cerevisiae allergic patients are advised to avoid yeast inhalation or eliminate yeast ingestion and preferably to have yeast-free diet.
Saccharomyces cerevisiae (S. cerevisiae) is also known as baker's yeast or brewer's yeast. It ferments sugar while producing carbon dioxide and ethanol (1). S. cerevisiae is a unicellular fungus, extensively utilized as a model organism during basic research (2).
It is an edible budding yeast that is considered safe and therefore possesses various applications in food, beverage, pharmaceuticals, and biofuels (3). Moreover, S. cerevisiae has a wide range of applications in both the food and beverage industry. This includes the baking industry (for bread making), the chocolate industry, the beverage industry (for the production of wine, beer, and cider), and distilled beverages (like rum, vodka, whisky, brandy) (2).
S. cerevisiae is a fungus naturally found in plants and soil. Environmental S. cerevisiae strains are found in soil, where they usually sporulate. They are prevalent in the leaves and trunk of trees and also in the environment of wineries (2). They can grow both aerobically and anaerobically, depending on the type of sugar. For aerobic growth, galactose and fructose are the preferred options for fermenting sugar. Every strain demands nitrogen, phosphorus, and sulfur, and different metals to ensure optimal development. The ideal growth condition that promotes yeast’s best morphological properties is found to be at a pH of 4 and with 5% dissolving oxygen (4).
Baker’s yeast belongs to the phylum Ascomycota that approximately contains 6,500 species. The fungi in this phylum show wide variation in habitat, pathogenicity, and size including unicellular yeast to large multicellular organism with fruiting bodies (1).
Taxonomic tree of S.cerevisiae (1) | |
---|---|
Domain | Eukaryota |
Kingdom | Fungi |
Phylum | Ascomycota |
Class | Saccharomycetes |
Order | Saccharomycetales |
Family | Saccharomycetaceae |
Genus | Saccharomyces |
Species | S. cerevisiae |
Taxonomic tree of S.cerevisiae (1) |
---|
S. cerevisiae is round-to-oval in shape and reproduces through budding mechanism. It grows in both haploid and diploid forms. It has a thick outer cell wall majorly composed of beta-glucan and a small amount of chitin along with a mannoprotein layer. This cell wall provides mechanical strength to the yeast cell (4).
The pervasiveness of allergenic sensitization to fungi is not known clearly; however, few studies claimed it to be around 6% in the general population (5).
Prevalence of type I allergy caused by Saccharomyces is found to be 1.4% in the general population and 7.4% in atopic individuals (6).
A study by Marszalska et al. (2001) aimed to show enolase (a major allergen from S. cerevisiae)-mediated immediate allergic response in patients with inhalant allergy sensitized to Candida albicans (C. albicans) extract. The study included a total of 70 patients divided into three groups (group I: 20 atopic patients with respiratory allergy sensitized to C. albicans and inhalant allergens such as mite, feather, pollens, group II: 30 patients with respiratory allergy, showing positive and negative skin tests to inhalant allergens and C. albicans respectively, group III: 20 nonatopic, healthy individuals) and detected 95% of group I patients and 10% of group II patients showing positive skin reactions to S. cerevisiae enolase (7).
Another study by Savolainen et al. (1993) in Finland evaluated the sensitizing capacity of S. cerevisiae in 449 allergic subjects [226 with atopic dermatitis (AD), 50 with allergic rhinitis (AR) and/or asthma, and 173 nonatopic controls], through skin prick tests (SPT). Out of 449 patients, 20% (92 patients; 85 with AD 4 with AR/asthma, and 3 nonatopic controls) showed positive SPT reaction to S. cerevisiae. (8).
Different strains of S. cerevisiae are found in various parts of the world. Wine strains are found in Europe, Australia, Chile, and New Zealand; oak strains are present in the woodlands of North America; and bertam palm strains in the Malaysian rainforests (9). In the northern hemisphere, Saccharomyces is commonly found in oak trees, while in the southern hemisphere, it is present in southern beech trees. Moreover, wild S. cerevisiae has been isolated from Chinese forests (10).
Inhalation of S. cerevisiae aeroallergen is considered as the primary route of exposure. (5, 11).
Another route of exposure to S. cerevisiae allergens is via ingestion (11).
In sensitized individuals, exposure to S. cerevisiae may lead to respiratory symptoms, such as asthma, particularly baker's asthma, AR, and hypersensitivity pneumonitis (1). Additionally, skin symptoms such as AD and chronic urticaria are reported to be caused by S. cerevisiae (7). Anaphylaxis is rarely reported due to baker’s yeast hypersensitivity (12).
Inhalation of fungal allergens may trigger rhinitis in sensitized individuals (5). A study by Baldo et al. (1988) evaluated 47 fungal allergic (inhalant allergen) patients for allergic sensitivity to bakers' yeast. As per the result, 32 patients were radioallergosorbant test (RAST)-positive to bakers' yeast antigens. Enolase was the major allergen identified from S. cerevisiae. (16).
Baker's asthma is a well-known occupational allergic condition that occurs due to hypersensitivity to different antigens, such as flour, bran, yeast, baking additives, storage mites, etc. Moreover, other ingredients found in baked goods and pastry have been reported to cause allergy (13). Baker’s yeast (S. cerevisae) is a bakery ingredient that has been reported to be associated with some isolated cases of baker’s asthma (14). However, sensitization to this baker’s yeast allergen is not common compared to the cereal flours or enzymes. Nonetheless, this needs to be considered in the clinical setting when no sensitization is observed for common bakery allergens (15).
Hernandez et al. (1996) presented a case of a 48-year-old bakery worker (nonsmoker) with recurrent episodes of hydrorrhea, sneezing, nasal obstruction, wheezing, spasmodic cough, and dyspnea within 1 or 2 hrs, following the start of his work. The symptoms at first were mild and worsened with time. However, his symptoms appeared to improve on holidays. The causative agent was found to be S. cerevisiae dry powder that created a dusty environment and possibly triggered occupational asthmatic condition in the baker (baker's asthma) (13).
Another case of baker's yeast true allergy in a boy with a history of mite-allergy and AD was reported by Pajno et al. (2005). The boy experienced generalized urticaria and asthma after eating pizza and bread (only fresh oven-baked). On investigation, he was found to be sensitized only to baker's yeast (S. cerevisiae), and the prick-by-prick procedure observed the severe systemic reaction. The severity of allergic reactions and urticaria subsided in 2 years. However, after the consumption of freshly baked and yeast-containing products, the patient still had a cough. The study suggested that continuous exposure to S. cerevisiae may result in immunotolerance and gradual reduction of allergic symptoms (11).
S. cerevisiae exhibited a significant association between positive SPT and AD (6).
A study on 449 allergic patients reported that about 50% suffered from AD, 11% from AR and/or asthma, and 38.5% were nonatopic individuals. The study detected that AD patients with chronic dermatitis had considerably higher SPT reactions to S. cerevisiae than AR/ asthma or nonatopic controls. Skin-specific IgE was detected for brewer's yeast in 94% of patients with severe AD, 76% with moderate AD, and 25% with mild AD or no history of AD. This indicated that the brewer's yeast had caused sensitization. (8)
Invasive infection
Saccharomyces organisms are reported as a prime contributor to invasive infections, particularly in immunocompromised or severely ill patients (17). S. cerevisiae has been frequently isolated from the vaginal flora of 0.9% – 5.8% of women. Moreover, S. cerevisiae is thought to cause symptoms similar to Candida vaginitis in 0.4% of patients. S. cerevisiae was also isolated from the throat (16%), stool (23%), urine (10%), and perineum (20%) of patients with hematologic disease. Similarly, a comprehensive review of published reports on invasive Saccharomyces infection by Angoulvant et al. (2005) detected 92 confirmed Saccharomyces invasive infection cases, which was clinically similar to invasive candidiasis. In most patients, the causative organism of fungemias was S. boulardii (51.3%), a subtype of S. cerevisiae (17).
Irritable bowel disease (IBD) and S. cerevisiae autoantibodies
Anti- S. cerevisiae antibody (ASCA), an antiglycan antibody against the cell wall surface of baker’s yeast, is commonly detected in the sera of patients with Crohn’s disease (CD) and ulcerative colitis (UC), the subtypes of irritable bowel disease (IBD). The bakery workers commonly develop CD (also called baker’s disease), and a diet low in baker’s yeast has been found to lower CD. This indicates the involvement of ASCA in CD pathogenesis (18). Although the exact cause of ASCA generation is not clear, it has been postulated that higher permeability in the small bowel of CD patients may result in higher exposure of reactive immune cells towards yeast antigens (typically found in the intestinal microbiota), leading to CD (19). In CD patients, an increased level of IgG and IgA ASCA has been reported. In fact, IgG ASCA is considered as a biomarker for IBD (positive rate 60-70% in CD patients and 10-15% in UC patients) (18).
Avoidance
Patients hypersensitive to S. cerevisiae should avoid inhalation of yeast or eliminate the ingestion of yeast in their diet and prefer a yeast-free diet (20).
To date, no allergenic molecule for S. cerevisiae has been identified and published officially by the World Health Organization/International Union of Immunological Studies Allergen Nomenclature Sub-Committee (WHO/IUIS) (21).
However, different studies mentioned different allergenic molecules of S. cerevisiae. Sac c enolase, a 46.8 kDa enolase, is considered as the major allergen (6). Some other allergens are Sac c MnSOD, a 25.7 kDa manganese-dependent superoxide dismutase protein, Sac c CyP, a cyclophilin (rotamase) protein, HSP 90, a heat shock protein of 90 kDa (6). Sac P2 protein, a ribosomal protein, was another allergen of S. cerevisiae, which is known to be a minor fungal allergen, also found in other fungal species such as Alternaria alternata, Aspergillus fumigatus, and Cladosporium herbarum (22). Moreover, baker’s yeast profilin Sac c profilin (PYF1) has been found to have low IgE binding ability and also less allergenicity (23).
Fungal enzymes can be potent occupational aeroallergens and may be important sources of fungal allergens (5). Allergen enzymes identified are Sac c Carboxypeptidase (a serine carboxypeptidase), Sac c Glucosidase (a glucosidase), Sac c Invertase, a fructofuranosidase (5)
A study by Horner et al. (2008) evaluated the IgE antibody reactivity of commercial fungal enzymes in sera of 20 mold-allergic patients. The study revealed that invertase from S. cerevisiae (baker's yeast) was the most reactive one, followed by glucosidase from S. cerevisiae (brewer's yeast). Moreover, S. cerevisiae enzymes (glucosidase, maltase, invertase, and invertase V) exhibited 100% sensitivity in detecting mold-allergic patients (5).
A heat-stable and soluble glycoprotein, gp200, with a molecular weight of 200 kDa, which is a part of the cell wall of S. cerevisiae, had been isolated. A correlation has been reported between IgA and IgG against gp200 and inflammation in Crohn's disease (24).
Enolase is considered to be a cross-reactive allergen among various fungal species, including S. cerevisiae, C. herbarum, A. alternata, C. albicans, etc. Allergens from S. cerevisiae, such as Sac c Cyp, Sac s MnSOD, are also reported to cross-react between different allergenic molecules of other fungal phyla. HSP90 (Heat shock protein) from S. cerevisiae is homologous to A. fumigatus allergen Asp f 12, an HSP90 protein (6). C. albicans and S. cerevisiae are cross-reactive due to homology among allergens; however, specific common allergens are yet to be identified (25).
Ingestion of yeast may induce allergy in mold-allergic patients (26). This was observed in a case study. A patient (29-year-old American woman) with a respiratory IgE sensitization to fungi (such as Alternaria alternata, Cladosporium herbarum, Aspergillus fumigatus, and Penicillium notatum) and S. cerevisiae was reported to develop several anaphylactic reactions after eating pasta along with yeast sauces consisting of cross-reacting fungal allergens (12). The patient was able to eat bakery products but developed allergic reactions for food or sauces with yeast flavoring. Moreover, increased exposure to mold aeroallergens leads to higher sensitivity towards the ingested yeast species. This indicated that yeast is likely to be an ingestive allergen in fungi sensitized persons (12).
Cross-reactivity was found among bakers' yeast, enolase from bakers' yeast, and C. albicans (16).
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Christian Fischer
Last reviewed: January 2021