Why choose Thermo Fisher Scientific as your partner for ANCA-associated vasculitis, Goodpasture syndrome and anti-GBM disease testing?
Best-in-class, fully automated testing for ANCA-associated vasculitis—putting the patient first.
EliA™ PR3S and EliA™ MPOS tests use innovative anchor technology to produce fully automated tests with the highest combination of specificity (98.0 % and 99.3%, respectively) and sensitivity (79.0% and 55.0%, respectively) on the market.6 Using the EliA™ ANCA-associated vasculitis portfolio enables your lab to help confidently differentiate diagnoses, and help ensure patients receive the correct treatment sooner.
Lean workflow
Patients with ANCA-associated vasculitis may have anti-GBM antibodies and the other way round, many patients with Goodpasture syndrome / anti-GBM disease may have antibodies against MPO-ANCA.4 Using EliA™ tests for these diseases enables you to benefit from the efficiencies of performing all three relevant antibody tests, from one sample, in one run, on the same fully automated Phadia™ Laboratory Systems. With the random-access functionality, you can add tests as they arrive, thus minimizing turnaround time. You can also save time and costs by utilizing the monthly calibration per method and sharing common system reagents.