Recent searches Clear History
Contacto
Allergen Encyclopedia
Table of Contents
Alérgeno completo
f283 Oregano
f283 Oregano Scientific Information
Tipo:
Whole Allergen
Nombre para mostrar:
Oregano
Route of Exposure:
Ingestion
Family:
Lamiaceae
Species:
Origanum vulgare
Latin Name:
Origanum vulgare
Other Names:
Spanish thyme, Wild marjoram
Summary
Oregano is a perennial aromatic plant frequently used as seasoning in Mediterranean countries, particularly in Turkish, Greek and Italian cooking. Food allergic reactions caused oregano are very rare, with occurrences being limited to single case reports. Symptoms of oregano allergy include pruritus, swelling of the lips and tongue and upper respiratory difficulty. Contact reactions to oregano are also possible in sensitized individuals.
Allergen
Nature
Oregano is a perennial aromatic plant belonging to the Lamiaceae family, which also contains basil, marjoram, mint, sage and lavender (1, 2). Oregano has been cultivated in the Mediterranean area for hundreds of years and now can be found worldwide. It is frequently used as culinary seasoning due to its distinctive taste, especially in Turkey, Greece and Italy. Oregano leaves and oil have been used for medicinal purposes in many cultures, where they believed to have antibiotic and antioxidant properties (3, 4). 
Taxonomy
Taxonomic tree of Olive (5)
Domain
Eukaryota
Kingdom
Plantae
Phylum
Tracheophyta
Subphylum
Spermatophytina
Class
Magnoliopsida
Family
Lamiaceae
Genus
Origanum
Epidemiology
Worldwide distribution 
Allergic reactions caused by ingestion of oregano are very rare, with occurrences being limited to single case reports (2). 
Route of Exposure
Main
Ingestion (2).
Clinical Relevance
Oregano allergy caused by ingestion has been reported in a 45-year-old male patient. He had previous history of IgE-dependent rhinitis and asthma. Within a few minutes of ingesting pizza containing oregano, the patient exhibited pruritus, swelling of the lips and tongue, dysphagia, dysphonia, progressive upper respiratory difficulty, and intense facial and palpebral edema. The symptoms abated after receiving epinephrine, corticosteroids, antihistamines and fluid therapy in an emergency care unit. Skin prick testing for was positive for oregano and thyme, as well as other members of the Lamiaceae family, reactions were negative in 10 control subjects. The patient subsequently experienced allergic reactions to food containing thyme alone. The patient had detectable specific IgE levels to oregano, thyme, marjoram and basil, the control subjects were negative (2). 
Atopic Dermatitis
In a study of 55 patients with suspected contact dermatitis, skin patch testing with a group of spices was performed, with the most common reactions being to ginger (12.7%), nutmeg (9.1%), and oregano (7.3%) (6).
Molecular Aspects
Allergenic molecules
The following allergens have been characterized from Origanum vulgare leaves (Allergome 2021):
Allergen
Type
Mass (kDa)
Ori v
Unknown function
-
Allergens present in oregano have not been well characterized (2). 
Cross-reactivity
Although reports are limited, cross-sensitivity between oregano and other plants of the Lamiaceae family (e.g. thyme, marjoram and basil) has been suggested (2).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer 
Last reviewed:April 2022
References
  1. Basavand E, Babaeizad V, Mirhosseini HA, Dehghan Niri M. Occurrence of leaf spot disease caused by Phoma herbarum on oregano in Iran. Journal of Plant Pathology. 2020;102(2):575-6.
  2. Benito M, Jorro G, Morales C, Peláez A, Fernández A. Labiatae Allergy: Systemic Reactions Due to Ingestion of Oregano and Thyme. Annals of Allergy, Asthma & Immunology. 1996;76(5):416-8.
  3. Singletary K. Oregano: Overview of the Literature on Health Benefits. Nutrition Today. 2010;45:129-38.
  4. Neill C, Nelssen J, Tokach M, Goodband R, Derouchey JM, Dritz S, et al. Effects of oregano oil on growth performance of nursery pigs. Journal of Swine Health and Production. 2006;14.
  5. ITIS. Origanum vulgare 2021 [cited 2021 18.01.22]. Available from: https://www.itis.gov/servlet/SingleRpt/SingleRpt?search_topic=TSN&search_value=32632#null.
  6. Futrell JM, Rietschel RL. Spice allergy evaluated by results of patch tests. Cutis. 1993;52(5):288-90.