Allergy to Fb was first observed in an occupational setting, where two plant leasing workers were sensitized and developed Fb-linked rhinitis and asthma. In a subsequent study in four plant-leasing firms, it was found that 27% of the workers had IgE antibodies to Fb. The sensitization occurred in atopic individuals as well as in non-atopic (1).
Since then, allergy to Fb has been reported to be around 3% among consecutive patients refered to a specialist for allergy investigation (2). These patients generally had a lower degree of exposure, and were sensitized in their homes or in public areas. Furthermore, non-atopic patients have been reported to develop allergic rhinoconjunctivitis, contact urticaria and asthma caused by Fb (3, 4).
Other topics
The genus Ficus includes over 1000 species. About ten of these, plus several varieties, are available as indoor plants.Ficus benjamina (Fb), also known as Java willow, Ceylon willow and Bali fig tree, is a tropical non-flowering plant used ornamentally in homes and public places such as offices and hospitals. It is decorative, easy to cultivate and it thrives indoors. The plant often grows into a small tree, reaching a height of 2-3 m.. A total of 11 allergenic components have been identified, three of them major allergens (5).
Allergy to Fb was first observed in an occupational setting, where two plant leasing workers were sensitized and developed Fb- linked rhinitis and asthma. In a subsequent study in four plant-leasing firms, it was found that 27% of the workers had IgE antibodies to Fb. The sensitization occurred in atopic individuals as well as in non-atopic (1).
Since then, allergy to Fb has been reported to be around 3% among consecutive patients refered to a specialist for allergy investigation (2). These patients generally had a lower degree of exposure and were sensitized in their homes or in public areas. Furthermore, non-atopic patients have been reported to develop allergic rhinoconjunctivitis, contact urticaria and asthma caused by Fb (3, 4). The main route for sensitization is probably by inhalation of airborne dust emanating from the leaves of the plant