Whole Allergen
f270 Ginger
f270 Ginger Scientific Information
Type:
Whole Allergen
Display Name:
Ginger
Allergen code:
f270
Family:
Zingiberaceae
Latin Name:
Zingiber officinale
Other Names:
Ginger, Ginger root, Green (Fresh) Ginger
WHO/ICD-11 code:
XM1TN8
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Clinical Relevance
IgE-mediated reactions
Sensitisation to Liliaceae has also been observed. Garlic, Onion or chive sensitivity was observed in 4.6% of skin prick tests in children, and in 7.7% in adults. No positive skin-specific IgE test occurred for Nutmeg, Ginger and Clove (1).
Skin-specific IgE tests with powdered commercial spices performed in 70 patients with positive skin-specific IgE test for Birch and/or Mugwort pollens and Celery yielded a report of positive tests in more than 24 patients for Anise, Fennel, Coriander or cumin—all from the same botanical family (Apiaceae) as Celery. Spices from unrelated families (Red Pepper, White Pepper, Ginger, Nutmeg, Cinnamon) elicited positive immediate skin test reactions in only 3 of 11 patients(2).
Allergic reactions have been reported to inhalation of Ginger dust (3).
Although tests for Ginger allergy have predominantly been for immediate hypersensitivity reactions, a greater prevalence of Ginger allergy is recorded using tests for delayed allergy reactions. Of 55 patients with suspected contact dermatitis, patch test results (positive at concentrations of 10% and 25%) were most common with Ginger(3) , Nutmeg (1), and Oregano (4); the remaining spices produced 0 or 1 positive responses (5).
Of about 1,000 patients investigated for occupational skin disease, 5 had occupational allergic contact dermatitis from spices. All patients had hand (or finger) dermatitis. The causative spices were Garlic, Cinnamon, Ginger, Allspice and Clove (6).
Other reactions
Anecdotally, excess Ginger ingestion has been said to cause irritation of the urethra.
As with a number of other common herbal remedies, such as Feverfew, Garlic, Ginkgo, and Asian ginseng, Ginger may increase the risk of bleeding during surgical procedures or potentiate the effects of warfarin therapy (7-9).
However, a study concluded that the effect of Ginger on thromboxane synthetase activity is dose-dependent, or only occurs with fresh Ginger, and that up to 2 g of dried Ginger is unlikely to cause platelet dysfunction when used therapeutically (10)
Molecular Aspects
No allergens from this plant have yet been characterized.
Cross-reactivity
An extensive cross-reactivity among the different individual species of the genus could be expected (4).
Compiled By
Last reviewed: April 2022
References
  1. Moneret-Vautrin DA, Morisset M, Lemerdy P, Croizier A, Kanny G. Food allergy and IgE sensitization caused by spices: CICBAA data (based on 589 cases of food allergy). Allergie et immunologie. 2002;34(4):135-40.
  2. Stäger J, Wüthrich B, Johansson SG. Spice allergy in celery-sensitive patients. Allergy. 1991;46(6):475-8.
  3. van Toorenenbergen AW, Dieges PH. Immunoglobulin E antibodies against coriander and other spices. The Journal of allergy and clinical immunology. 1985;76(3):477-81.
  4. L. Y. Botanical relations and immunological cross-reactions in pollen allergy. 2nd ed Pharmacia Diagnostics AB Uppsala Sweden. 1982.
  5. Futrell JM, Rietschel RL. Spice allergy evaluated by results of patch tests. Cutis. 1993;52(5):288-90.
  6. Kanerva L, Estlander T, Jolanki R. Occupational allergic contact dermatitis from spices. Contact dermatitis. 1996;35(3):157-62.
  7. Fessenden JM, Wittenborn W, Clarke L. Gingko biloba: a case report of herbal medicine and bleeding postoperatively from a laparoscopic cholecystectomy. The American surgeon. 2001;67(1):33-5.
  8. Heck AM, DeWitt BA, Lukes AL. Potential interactions between alternative therapies and warfarin. American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists. 2000;57(13):1221-7; quiz 8-30.
  9. Pribitkin ED, Boger G. Herbal therapy: what every facial plastic surgeon must know. Archives of facial plastic surgery. 2001;3(2):127-32.
  10. Lumb AB. Effect of dried ginger on human platelet function. Thrombosis and haemostasis. 1994;71(1):110-1.