Allergen Exposure
Geographical distribution
Potentially allergenic molds form a broadly distributed group of organisms. They exist almost worldwide, and are found in a great variety of environments, since they are adapted, and continue to adapt, to make use of a wide spectrum of food sources, which as microorganisms with airborne spores they can infiltrate easily and unobtrusively. Of greatest concern for possible allergic sensitisation is the prominence of these molds in domestic and work environments and in infections of humans. In this latter capacity we can characterise potentially allergenic molds most distinctly, as they have been fairly extensively studied as infectious agents, but hardly at all as allergens.
The genus Trichosporon consists of non- or weakly fermentative fungi. There is no sexual reproduction phase. Their primary mode of reproduction is the formation of arthrospores and blastospores. Trichosporon colonies are yeast-like, rapidly growing, wrinkled, raised, folded, glabrous to velvety, and white to yellowish-white. The wrinkles become more prominent in time. Heaping at the centre of the colony is typical. Urease enzyme production is a significant feature of this genus (1).
A rearrangement of the genus has been proposed as a result of the use of modern molecular biology study (2-3). The newly proposed nomenclature holds that there are 17 species, of which 6 are associated with infections in humans (2). This nomenclature has not yet been fully accepted.
The species causing human diseases are said to be the following:
- T. cutaneum syn. T. beigelii
- T. asteroids
- T. ovoides
- T. inkin
- T. asahii
- T. mucoides
Other species of Trichosporon are T. brassicae, T. coremiformis, T. dulcitum, T. faecalis, T. gracile, T. jirovecii, T. lowideri, T. montevidense, T. muwides, T. paehachi, T. pullulans, and T. sporotrichoides.
Trichosporon spp. are the causative agents of white piedra (superficial infections of hair shafts), onychomycosis (infections of the nails), other superficial infections, and invasive trichosporonosis. In recent years, this fungus has emerged as an opportunistic fungal pathogen. Immunocompromised hosts are particularly at risk of developing invasive infections (4). Cutaneous lesions as a manifestation of disseminated infection are also likely in these patients. Trichosporon is one of the fungi isolated from patients with fungemia. Trichosporon frequently induces summer-type hypersensitivity pneumonitis, which is the most prevalent type of hypersensitivity pneumonitis in Japan (5).
Trichosporon beigelii is a minor component of normal skin flora, and is widely distributed in nature, but nevertheless is the most significant pathogen in the genus Trichosporon. (The name Trichosporon cutaneum is often used synonymously with Trichosporon beigelii.) T. beigelii causes white piedra, onychomycosis, otomycosis, and superficial skin infections. This organism is also associated with the summer-type hypersensitivity pneumonitis commonly found in Japan (6).
T. inkin and T. ovoides are associated with white piedra (T. ovoides with cranial white piedra and T. inkin with pubic white piedra). T. asteroides and T. cutaneum are associated with superficial skin lesions. T. asahii and T. mucoides are associated with systemic infection and disseminated trichosporonosis (7)
Environment
Trichosporon is a cosmopolitan yeast, common in soil, fresh and sea water, vegetables, mammals and birds. It has also been isolated from garden compost, sewage sludge, inter-tidal sand, and mud. It is a common coloniser of human skin and GI tracts. As well as being a member of the normal flora of mouth, skin and nails, it is the causative agent of superficial and deep infections in humans. (See above under Geographical distribution.)