Whole Allergen
t55 Scotch broom
t55 Scotch broom Scientific Information
Type:
Whole Allergen
Display Name:
Scotch broom
Allergen code:
t55
Family:
Fabaceae (Leguminosae)
Source Material:
Pollen
Latin Name:
Cytisus scoparius
Other Names:
Scotch broom, Common broom, English broom, Broom tops, Irish tops, Basam, Bisom, Bizzom, Browme, Brum, Breeam, Green broom
Environmental Characteristics
Worldwide distribution 
Scotch broom is a perennial, leguminous shrub native to western and central Europe, from the Iberian Peninsula north to the British Isles and southern Scandinavia, and east to Poland and Romania. It has been introduced into several other continents outside its native range and is classified as a noxious invasive species in many countries, including the US (on the East Coast and in the Pacific Northwest), South Africa, Australia and New Zealand. It typically grows to 1-3 m tall, with main stems up to 5 cm thick. It has green shoots with small deciduous trifoliate leaves 5-15 mm long. Its long, slender, erect and tough branches grow in large, close fascicles, making it apt for broom-making.(1)
In spring and summer, it is covered in profuse golden-yellow flowers 20-30 mm long and 15-20 mm wide. In late summer, its seed pods mature, becoming black, 2-3 cm long, 8 mm broad and 2-3 mm thick; they burst open, often with an audible crack, spreading seed.
There are 2 subspecies. Scotch broom is widely cultivated as an ornamental plant.
The twigs and branches are used for making brooms and also for basket-work. Parts of the plant have been employed medicinally, and are thought to be diuretic and cathartic
Clinical Relevance
IgE-mediated reactions
Anecdotal evidence suggests that asthma and hayfever are possible following exposure to pollen from this plant; however, no studies have been reported to date.
Other reactions
Attempts have been made to develop biological controls in affected areas, using Broom-feeding insects, e.g., the psyllid Arytainilla spartiophylla, the beetle Bruchidius villosus, and the moth Leucoptera spartifoliella. Since allergic reactions have been reported to these insects, they should be considered as potential allergens in individuals thought to be allergic to pollen from this plant, but in whom specific investigations are negative.
Molecular Aspects
No allergens have been characterized.
Compiled By
Last Reviewed : June 2022
References
  1. Weber RW. On the Cover. Annals of Allergy, Asthma & Immunology. 2009;103(3):A4.