Rubottom oxidation reaction
In 1974, G.M. Rubottom, A.G. Brook, and A. Hassner independently created a method to prepare alpha-hydroxy aldehydes and ketones through the oxidation of their silyl enol ethers using meta-chloro peroxy benzoic acid (mCPBA). Today, the peroxyacid oxidation of silyl enol ether substrates to prepare the corresponding alpha-hydroxy carbonyl compounds is known as the Rubottom oxidation. Synthesis of the potent anti-thrombotic (+/-)rishirilide B utilized the Rubottom oxidation, as did the synthesis of the antitumor antibiotic FR901464.
Review available Thermo Scientific products for the Rubottom oxidation reaction:
Catalog # Name Size Price (USD) Qty
L00286.14 3-Chloroperoxybenzoic acid, 50-55%, cont. ca 10% 3-chlorobenzoic acid, balance water Each -
L00286.22 3-Chloroperoxybenzoic acid, 50-55%, cont. ca 10% 3-chlorobenzoic acid, balance water Each -
L00286.36 3-Chloroperoxybenzoic acid, 50-55%, cont. ca 10% 3-chlorobenzoic acid, balance water Each -
A12535.09 Trimethylsilyl trifluoromethanesulfonate, 99% Each
39.65
Special offer
Online exclusive
Ends: 30-Jun-2025
46.30
Save 6.65 (14%)
A12535.18 Trimethylsilyl trifluoromethanesulfonate, 99% Each
126.65
Special offer
Online exclusive
Ends: 26-Sep-2025
149.00
Save 22.35 (15%)
A12535.30 Trimethylsilyl trifluoromethanesulfonate, 99% Each
473.65
Special offer
Online exclusive
Ends: 26-Sep-2025
557.00
Save 83.35 (15%)
A12646.AK Triethylamine, 99% Each
38.60
A12646.AP Triethylamine, 99% Each
47.90
A12646.0F Triethylamine, 99% Each
116.65
Special offer
Online exclusive
Ends: 26-Sep-2025
137.00
Save 20.35 (15%)
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote now