Accurate, sensitive, and reliable real-time PCR system designed for the clinical laboratory
The Applied Biosystems QuantStudio 5 Dx Real-Time PCR System is designed to streamline workflows and minimize training needs, helping you get clinical answers quickly by integrating seamlessly into your established workflow.
Video Player is loading.
Current Time 0:00
Duration 0:36
Loaded: 0.00%
Stream Type LIVE
Remaining Time 0:36
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Customer testimonial
    “Thermo Fisher is a valued partner, supporting us in the development of clinical trial assays and companion diagnostics,” said Michael Sloan, global vice president of commercial operations at Almac Diagnostic Services. “Thermo Fisher’s QuantStudio qPCR systems offers a seamless integration into our assay development workflows. QuantStudio 5 Dx offers the flexibility of customizable solutions and provide high-quality data output for molecular diagnostic testing.”
    Michael Sloan
    Global Vice President of Commercial Operations
    Almac Diagnostic Services
    Features
    Features of the QuantStudio 5 Dx Real-Time PCR System include:
    Results you can trust
    Detect differences in target quantity as small as 1.5-fold in singleplex reactions and obtain 10 logarithmic units of linear dynamic range
    Designed with the clinic in mind
    An interactive diagnostic instrument with short run times and minimal maintenance that also uses existing plastic consumables for added convenience in clinical laboratories
    Established system
    Thermo Fisher Scientific has over 10 years of experience manufacturing clinical real-time PCR instruments
    Enhanced security
    Security, auditing and e-signature functionalities that enable you to comply with regulations, including 21 CFR Part 11
    Simple, powerful software
    A single, easy-to-use software interface allows users to set up a run, lay out assays, control the instrument, and conduct plate analysis
    IVD Test mode
    The in vitro diagnostic (IVD) test menu allows only authorized IVD tests to be run, reducing the risk of unauthorized use and accidental or intentional misuse
    Highly structured or highly flexible
    Diagnose or develop, the choice is yours with software options that guide you through every step of test development* and IVD modes
    QuantStudio performance
    High reliability, sensitivity, and accuracy with an intuitive and simple-to-use interface that allows users of any experience level to operate the system with ease
    Global support
    Support by highly-skilled, customer-focused scientists available globally
    Maximize benchtop space
    Compact instrument preserves space for other equipment
    Features that help you comply with requirements of accrediting bodies
    Maintenance and calibration reports
    Records are updated automatically with maintenance and calibration events and can be printed on demand, documenting that the system has been maintained and calibrated to vendor specifications.
    Reagent tracking
    Information about the reagents used with each test run, including lot number and expiration date, is stored and archived. Archived files can be retrieved when required to track samples that were tested with a given set of reagents.
    Sample tracking
    The system enables laboratories to more easily track samples associated with a particular plate, set of reagents, run date and time, and data files.
    E-signature history
    Security, auditing, and e-signature software records test events, actions taken, dates, usernames, user roles, and the activity performed for documentation and archiving purposes.
    Experimental results
    For documentation, archiving, and review-at-a-glance needs, detailed reports include experiment name, barcode, file name, time stamps (creation, run start, run finish, duration, and modifications), instrument name, serial number, experiment type, results summary, plate layout image, standard curves, results table, and QC summary.
    Specifications
    Sample capacity
    96
    Reaction volume
    96-well 0.2-mL block: 10–100 μL
    Excitation source
    Bright white LED
    Filter or color combinations
    6 decoupled filters, CMOS camera
    Excitation/detection range
    450–680 nm/500–730 nm
    Multiplexing
    5-plex + 1 passive reference, 6-plex with no passive reference
    Maximum block ramp rate
    6.5°C/sec
    Average sample ramp rate
    3.66°C/sec
    Temperature uniformity
    0.4°C
    Temperature range
    4–99.9°C
    Heating and cooling method
    Peltier
    Independent temperature zones
    6 VeriFlex zones (5°C zone to zone)
    Chemistries
    Fast and standard
    Run time
    <30 min
    Compatible dyes
    FAM, SYBR Green, VIC, ABY, NED, TAMRA, JUN, ROX, Mustang Purple, Cy5
    Throughput
    Medium
    Thermal block style
    Fixed
    Thermal block format
    96-well (0.2-mL block)
    Attached computer
    Required
    Automation capable
    No
    Instrument size
    40 x 27 x 50 cm; 27 kg
    Touchscreen size
    8.5 inch
    Multiplexing and filters
    Up to 6 targets; 21 filter combinations
    Hands-free functionality
    No
    Smart Help
    No
    Security, audit, and e-signature functionality
    The audit function is always enabled in IVD mode and on the desktop software when in Development mode*.   In Development mode, audit mode can be set to Optional or Required depending on traceability needs.
    Ordering information
    Reagents
    Applied Biosystems TaqPath real-time PCR master mixes deliver confidence and reliability for your most demanding molecular diagnostic development and testing needs. Application-specific formulations of TaqPath master mixes are available for qPCR and one-step RT-qPCR using 5-prime nuclease assays in singleplex or multiplex format. Each reagent is manufactured and labeled in accordance with requirements for general-purpose reagents and is functionally tested to help ensure lot-to-lot reproducibility for Cq consistency and a wide dynamic range.
    *Development mode is For Research Use Only. Not for use in diagnostic procedures.
    For In Vitro Diagnostic Use.