c7c8 - Key/Specs/Resources/Appl/Tech/Doc/Contact
Demand for high-performance, energy-efficient electronics is driving the development of advanced devices with ever smaller, more densely packed features and complex 3D structures. Ramping production of these cutting-edge microprocessors, memory devices, and other products is extremely challenging and requires high-resolution, atomic-scale analysis of features buried deep within the device. Transmission electron microscopy (TEM) is increasingly becoming the go-to technique for this kind of analysis and relies on high-quality samples produced with focused ion beam (FIB) milling.
The Thermo Scientific Helios 5 EXL DualBeam is a 300mm full-wafer focused ion beam scanning electron microscope (FIB-SEM), designed to address TEM sample preparation challenges in the semiconductor industry. The Helios 5 EXL DualBeam is capable of preparing samples for today’s most advanced process nodes, including sub-5nm and gate-all-around technology.
Maximizing sample throughput and productivity
Utilizing advanced machine learning and closed-loop end pointing, the Helios 5 EXL DualBeam delivers enhanced cut placement precision and enables you to consistently extract high-quality lamella from your most challenging samples.
Flexible automation capabilities make ultra-thin TEM sample generation routine and consistent, providing unparalleled, sub-nanometer insight into more interfaces, films, and profiles to measure at sub-nanometer resolution. The Helios 5 EXL DualBeam ensures efficient and consistent TEM sample preparation workflows by combining wafer and defect navigation with recipe definition and execution in a single, fully integrated program. This automation supports a higher tool-to-operator ratio, maximizing sample throughput and technical resource productivity.
Video Player is loading.
Current Time 0:00
Duration 2:55
Loaded: 0.00%
Stream Type LIVE
Remaining Time 2:55
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Revolutionizing Semiconductor Analysis: Thermo Scientific Helios 5 EXL Overview
    TEM image of a gate-all-around transistor.
    TEM image of a Si/SiGe gate-all-around (GAA) fin.
    TEM cross-section of a transistor gate along a nanowire.
    TEM image of a gate cross-section along a nanowire.
    Key Features
    Automated TEM sample preparation software
    Thermo Scientific AutoTEM 5 Software combines wafer and defect navigation with recipe definition and execution in a single, fully integrated program, ensuring efficiency and consistency among operators with varying levels of expertise. AutoTEM Software simplifies TEM sample preparation, allowing users to easily schedule multi-site jobs for inverted, plan-view, and top-down TEM sample preparation workflows.
    Repeatable, automated deposition and etch
    Developed specifically to support automated TEM preparation, the Thermo Scientific MultiChem Gas Delivery System provides highly consistent deposition and etching capabilities and can be utilized with automated applications. The motorized injection needle with saved position presets can be accurately positioned for optimized, reproducible gas delivery to the sample surface. The MultiChem Gas Delivery System is also engineered for serviceability, maximizing tool uptime.
    Precision FIB milling for advanced sample preparation
    The Helios 5 EXL DualBeam includes the Thermo Scientific Phoenix Ion Column, which provides revolutionary low-kV performance and leading-edge TEM sample preparation.
    Automated alignments, high resolution, and consistent results
    The high-performance Thermo Scientific Elstar Electron Column features our unique UC monochromatic technology, offering improved resolution and TEM sample end-pointing. New SEM auto-alignments ensure consistent results across multiple tools and operators.
    Automated sample manipulation and lift out
    With an intuitive method for lift-out and transfer of TEM samples to a grid, the Thermo Scientific EasyLift Nanomanipulator offers low-drift, high-precision movements for simple and consistent creation of traditional or ultra-thin TEM lamella. Highly accurate, easy-to-use, and fast motorized rotation makes the EasyLift Nanomanipulator ideally suited for high-speed inverted or plan-view sample preparation.
    Fab-compatible Automated FOUP Loader (AFL) option
    The optional Automated FOUP Loader (AFL) enables the Helios 5 EXL DualBeam to be located inside the semiconductor wafer fab. By being closer to the wafer process line (near-line), it can deliver critical information up to three times faster than laboratory-based analysis of cleaved wafer pieces, accelerating the development of new processes and the yield ramp to high-volume production.
    Specifications
    Thermo Scientific Phoenix Ion Column
    • Gallium focused ion beam
    • Up to 65 nA maximum beam current
    • Low-voltage (500 V) performance for high sample preparation quality
    • Ion beam resolution at coincident point and 30 kV: 4.0 nm using preferred statistical method
    • Ion source lifetime: 1,000 hours guaranteed
    Thermo Scientific Elstar Electron Column
    • An ultra-high-resolution immersion-lens field-emission SEM (FESEM) column
    • The ultra-stable Schottky field emitter gun features UC+ monochromator technology
    • Electron beam resolution:
      • 1.0 nm @ 15 kV
      • 0.9 nm @ 1 kV
    • Electron source lifetime: 12 months
    Gas delivery
    • Thermo Scientific MultiChem Gas Delivery System
    • Slots for up to 6 individual chemistries
    • Single gas injection system
    • Port for up to 3 independent GIS units
    Detectors
    • Elstar in-lens SE detector (TLD-SE)
    • Elstar in-lens BSE detector (TLD-BSE)
    • High-performance ion conversion and electron (ICE) detector for secondary ions (SI) and electrons (SE)
    Sample handling
    • Automated handling of 300 mm FOUP with EFEM (GEM300 compliant)
    • Manual loading of 300 mm, 200 mm, and 150 mm wafers
    Additional options
    • Optical microscope with 920 µm field of view
    • 30 kV STEM detector with BF/DF/HAADF segments
    • Oxford EDS
    • CAD Navigation Compatibility (NEXS and Synopsys Camelot)
    • Thermo Scientific iFast Software Semiconductor Wafer Navigation (optional)
    • Integrated defect navigation based on KLARF 1.2 and 1.8 standards
    • User-defined wafer maps and site plans
    Resources
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        The Thermo Scientific Helios DualBeam Family
        Video Player is loading.
        Current Time 0:00
        Duration 2:09
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 2:09
        1x
          • Chapters
          • descriptions off, selected
          • en (Main), selected
          The Thermo Scientific Helios DualBeam Family
          Applications
          Semiconductor Device Packaging
          Semiconductor Advanced Packaging
          Performance, power efficiency, area, and cost are driving packaging innovations. Learn how workflows provide fast, precise, and accurate time-to-data.
          Semiconductor-RD_1920x1080
          Semiconductor research and development
          Innovation starts with research and development. Learn more about solutions to help you understand innovative structures and materials at the atomic level.
          Semi-Metrology_1920x1080
          Semiconductor metrology
          Manufacturing today’s complex semiconductors requires exact process controls. Learn more about advanced metrology and analysis solutions to accelerate yield learnings.
          Semiconductor Failure Analysis
          Semiconductor Failure Analysis
          Complex semiconductor device structures result in more places for defects to hide. Learn more about failure analysis solutions to isolate, analyze, and repair defects.
          Semi-Materials-Character_1920x1080
          Semiconductor materials characterization
          Many factors impact yield, performance, and reliability. Learn more about solutions to characterize physical, structural, and chemical properties.
          Techniques
          Semiconductor TEM Imaging and Analysis
          Thermo Scientific transmission electron microscopes offer high-resolution imaging and analysis of semiconductor devices, enabling manufacturers to calibrate toolsets, diagnose failure mechanisms, and optimize overall process yields.
          Learn more ›
          TEM Metrology
          Advanced and automated TEM metrology routines deliver significantly greater precision than manual methods. This allows users to generate large amounts of statistically relevant data, with sub-angstrom-level specificity, that is free of operator bias.
          Learn more ›
          Sample Preparation of Semiconductor Devices
          Thermo Scientific DualBeam systems provide accurate TEM sample preparation for atomic-scale analysis of semiconductor devices. Automation and advanced machine learning technologies produce high-quality samples, at the correct location, and a low cost per sample.
          Learn more ›
          Semiconductor TEM Imaging and Analysis
          Thermo Scientific transmission electron microscopes offer high-resolution imaging and analysis of semiconductor devices, enabling manufacturers to calibrate toolsets, diagnose failure mechanisms, and optimize overall process yields.
          Learn more ›
          TEM Metrology
          Advanced and automated TEM metrology routines deliver significantly greater precision than manual methods. This allows users to generate large amounts of statistically relevant data, with sub-angstrom-level specificity, that is free of operator bias.
          Learn more ›
          Sample Preparation of Semiconductor Devices
          Thermo Scientific DualBeam systems provide accurate TEM sample preparation for atomic-scale analysis of semiconductor devices. Automation and advanced machine learning technologies produce high-quality samples, at the correct location, and a low cost per sample.
          Learn more ›
          Contact us
          Electron microscopy services for
          semiconductors
          To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
          Learn more ›
          Style Sheet to change Applications H3 to p with em-h3-header class
          Style Sheet to change H3 to p with em-h3-header class
          Style Sheet to change H2 style to p with em-h2-header class
          Style Sheet for Komodo Tabs
          Style Sheet for Support and Service footer
          Style Sheet for Fonts
          Style Sheet for Cards