Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Acrylamide Gel Electrophoresis for DNA |
Polyacrylamide gel electrophoresis (PAGE) is a technique widely used in biochemistry, molecular biology, and genetics to separate macromolecules, such as proteins or nucleic acids, based on their size and charge. PAGE involves the use of a polyacrylamide gel matrix, which serves as a medium through which the molecules can migrate when an electric field is applied. PAGE provides high resolution of DNA molecules between 10–3,000 bp long. Under optimal conditions, DNA molecules differing in single nucleotide can be resolved using polyacrylamide gels.
You can rely on our reagents, including UltraPure acrylamide, for pouring your own gels and hassle-free precast Invitrogen Novex polyacrylamide gels when running PAGE gels.
We offer a wide variety of precast polyacrylamide Novex gels that can be used for standard or denaturing nucleic acid electrophoresis. To obtain the best result based on your application (PCR fragment separation, DNA retardation, etc.), it is important to choose the correct gel percentage, buffer system, and gel format. Figure 1 shows the migration pattern of DNA run on Novex polyacrylamide gels. Use the infographic to help select the optimal gel for your experiments.
The size of Novex precast gels is 10 x 10 cm (gel size is 8 x 8 cm). We recommend using the XCell SureLock Mini-Cell Electrophoresis System to obtain optimal and consistent performance.
Novex TBE (Tris-Borate-EDTA) gels provide the highest resolution for DNA fragments 10–3,000 bp long. Use these gels for any DNA electrophoresis application, including analyzing restriction-enzyme digests, PCR products, southern analysis, and primer analysis.
Novex TBE Gels, 4–20% |
| Order here |
Novex TBE Gels, 6% |
| Order here |
Novex TBE Gels, 8% |
| Order here |
Novex TBE Gels, 10% |
| Order here |
Novex TBE Gels, 20% |
| Order here |
Novex DNA Retardation Gels consist of 6% polyacrylamide prepared with 0.5X TBE as the gel buffer. They provide good resolution of 60–2,500 bp DNA fragments. 0.5X TBE buffer offers good fragment separation in electrophoresis, yet its ionic strength is low enough to promote DNA-protein interactions.
DNA Retardation Gels (6%), 1.0 mm |
| Order here |
Novex TBE-Urea Gels are optimized for analysis and purification of single-stranded DNA or RNA 20–800 bases long. They are an excellent choice for synthetic oligonucleotide analysis and purification, RNase protection assays, in vitro transcription studies, and northern blot analysis.
![]() Novex TBE-Urea Gels, 6% |
| Order here |
![]() Novex TBE-Urea Gels, 10% |
| Order here |
![]() Novex TBE-Urea Gels, 15% |
| Order here |
Novex TBE (Tris-Borate-EDTA) gels provide the highest resolution for DNA fragments 10–3,000 bp long. Use these gels for any DNA electrophoresis application, including analyzing restriction-enzyme digests, PCR products, southern analysis, and primer analysis.
Novex TBE Gels, 4–20% |
| Order here |
Novex TBE Gels, 6% |
| Order here |
Novex TBE Gels, 8% |
| Order here |
Novex TBE Gels, 10% |
| Order here |
Novex TBE Gels, 20% |
| Order here |
Novex DNA Retardation Gels consist of 6% polyacrylamide prepared with 0.5X TBE as the gel buffer. They provide good resolution of 60–2,500 bp DNA fragments. 0.5X TBE buffer offers good fragment separation in electrophoresis, yet its ionic strength is low enough to promote DNA-protein interactions.
DNA Retardation Gels (6%), 1.0 mm |
| Order here |
Novex TBE-Urea Gels are optimized for analysis and purification of single-stranded DNA or RNA 20–800 bases long. They are an excellent choice for synthetic oligonucleotide analysis and purification, RNase protection assays, in vitro transcription studies, and northern blot analysis.
![]() Novex TBE-Urea Gels, 6% |
| Order here |
![]() Novex TBE-Urea Gels, 10% |
| Order here |
![]() Novex TBE-Urea Gels, 15% |
| Order here |
Access learning resources—a knowledge base for all things nucleic acid electrophoresis.
View our webinars to learn more about molecular biology topics.
E-Gel Precast Gel and Ladder Selection Tool for Invitrogen Power Snap devices.
Find answers to your questions on nucleic acid gel electrophoresis using acrylamide or agarose gels, including E-gels.
For Research Use Only. Not for use in diagnostic procedures.