Search Thermo Fisher Scientific
Nalgene Pans, Trays and Baskets are made rugged with the rigors of lab life in mind. Nalgene pans have rolled rims and heavy wall construction to hold a full load of labware and other items to be moved about the lab. Use them for everything from dish pans to secondary containment for chemicals. Choose from an assortment of polypropylene pans, trays and baskets for autoclaving and sterilizing labware and instruments. Plastic construction is quieter than metal pans, and doesn't dent or corrode. Polypropylene and polyethylene pans are compatible with a wide variety of lab reagents and cleaning solutions including most acids, bases and alcohols.
Nalgene Large Sterilizing Pan | Nalgene Autoclave Pan | Nalgene Instrument/Pipet Sterilizing Tray with Cover | Nalgene Autoclave Basket |
Sink to autoclave; useful for collecting soiled labware and instruments for washing and autoclaving. Rugged construction. Molded-in handles for easy carrying. | Sink to autoclave; useful for collecting soiled labware and instruments for washing and autoclaving. Rugged construction. Rounded corners for easy cleaning. | Horizontal design recommended for soaking pipets and lab instruments in disinfectant. Meets OSHA and other regulatory requirements for pathogen safety. | Recommended for holding small lab items during autoclaving. Provides complete venting and draining. |
![]() | ![]() | ![]() | ![]() |
Polypropylene (PP) | Polypropylene (PP) | Filled Polypropylene | Polypropylene (PP) |
Autoclavable | Autoclavable | Autoclavable | Autoclavable |
2 sizes | 4 sizes | 1 size | 3 sizes |
Approx dims - English 14" x 12" x 6" (10 Qt) 21" x 17" x 5" (22 Qt) | Approx dims - English 10" x 6" x 2" (2 Qt) 10" x 6" x 4" (3 Qt) 13" x 10" x 3" (3 Qt) 13" x 10" x 4" (5 Qt) | Approx dims - English 18" x 6" x 3" | Approx dims - English 6" x 5" x 4" 6" x 7" x 7" 9" x 9" x 9" |
Approx dims - Metric 36 x 31 x 14cm (9.5L) 54 x 44 x 13cm (20L) | Approx dims - Metric 26 x 16 x 6cm (2L) 26 x 16 x 10cm (3L) 32 x 26 x 7cm (3L) 32 x 26 x 11cm (5L) | Approx dims - Metric 45 x 15 x 7cm | Approx dims - Metric 16 x 12 x 10cm 16 x 17 x 18cm 23 x 23 x 24cm |
Molded handle | No handle | No handle | No handle |
No cover | No cover | Cover fits in pan groove | Hinged cover |
Nestable for storage | Nestable for storage | Nestable for storage | Nestable for storage |
No compliance claims | No compliance claims | Meets OSHA 29 CFR 1910.1030 bloodborne pathogen protection, NIH and Fed Reg biohazard guidelines, and more | No compliance claims |
Nalgene Large Sterilizing Pan | Nalgene Autoclave Pan | Nalgene Instrument/Pipet Sterilizing Tray with Cover | Nalgene Autoclave Basket | |
Acids, dilute or weak | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G |
Alcohols, aliphatic | E | E | E | E |
Aldehydes | G | G | G | G |
Bases/alkali | E | E | E | E |
Esters | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N |
Ketones, aromatic | N | N | N | N |
Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20°C. |
G | Little or no damage after 30 days constant exposure at 20°C. |
F | Some effect after 7 days constant exposure at 20°C. |
N | Not recommended. Immediate damage may occur. |
Note: This pan is NOT autoclavable
For help selecting the Nalgene containers that are perfect for your lab applications, request a copy of our chemical compatibility wall chart, or for personal assistance, contact the Nalgene Technical Support.
Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Thermo Scientific Nalgene and Nunc products.
Need technical support? Contact our expert team for technical and application support of Laboratory Products at technicalsupport@thermofisher.com.
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.