Search Thermo Fisher Scientific
Nalgene buckets, pails and storage tubs come in a variety of configurations, but all have one thing in common - excellent chemical compatibility with a wide range of laboratory chemicals and production materials. These products are made with work in mind.
Nalgene Graduated HDPE Bucket | Nalgene Autoclavable Graduated Bucket | Nalgene LDPE Bucket with Lid | Nalgene Large Cylinderical HDPE Tub with Cover |
Super-rugged and chemical resistant, graduated | Fully autoclavable, graduated | Air-tight cover meets OSHA standards for bloodborne pathogens, graduated | Positive-lock cover provides a tight seal for transport or storage. Easy-grip side handles add comfort and durability for lifting and storing. |
![]() | ![]() | ![]() | ![]() |
High Density Polyethylene (HDPE) | Polypropylene (PP) | Low Density Polyethylene (LDPE) | High Density Polyethylene (HDPE) |
Polyethylene coated wire bail | Nylon-coated wire bail | Nylon-coated wire bail | Molded-in hand grips |
Graduated | Graduated | Graduated | No graduations |
Do not autoclave | Autoclavable | Do not autoclave | Do not autoclave |
Pour Spout | No pour spout | No pour spout | No pour spout |
No cover | No cover | Air-tight cover meets OSHA 29 CFR 1910.1030 for bloodborne pathogens | Positive-lock cover. NSF approved for food and beverage storage. |
Nalgene Graduated HDPE Bucket | Nalgene Autoclavable Graduated Bucket | Nalgene LDPE Bucket with Lid | Nalgene Large Cylinderical HDPE Tub with Cover | |
Acids, dilute or weak | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G |
Alcohols, aliphatic | E | E | E | E |
Aldehydes | G | G | G | G |
Bases/alkali | E | E | E | E |
Esters | G | G | G | G |
Hydrocarbons, aliphatic | G | G | F | G |
Hydrocarbons, aromatic | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N |
Ketones, aromatic | N | N | N | N |
Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20°C. |
G | Little or no damage after 30 days constant exposure at 20°C. |
F | Some effect after 7 days constant exposure at 20°C. |
N | Not recommended. Immediate damage may occur. |
For help selecting the Nalgene containers that are perfect for your lab applications, request a copy of our chemical compatibility wall chart, or for personal assistance, contact the Nalgene Technical Support.
Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Thermo Scientific Nalgene and Nunc products.
Need technical support? Contact our expert team for technical and application support of Laboratory Products at technicalsupport@thermofisher.com.
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.