Organism TypePrimate - Non HumanBSC-40 cells were a continuous line of African green monkey cells derived from BSC-1 cells (Hruby, et al., 1979). Cells were maintained as monolayer cultures in Dulbecco's modified Eagle's medium supplemented with 10% heat-inactivated fetal bovine serum and antibiotics.
TissuesKidney
PhenotypeAdherent
Primaryno
ApplicationCell Culture / Growth Conditions,Transient Transfection
Products
Catalog # Name Size Price (USD) Qty
11095098 MEM, 10 x 500 mL Each
300.65
Online Exclusive
324.00
Save 23.35 (7%)
18292011 Lipofectin™ Transfection Reagent, 1 mL Each
710.00
Every step of the way, a wide range of cell health products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.