Cytoskeleton of a mixed population of granule neurons and glial cells
Confocal micrograph of the cytoskeleton of a mixed population of granule neurons and glial cells. The F-actin was stained with red-fluorescent Texas Red®-X phalloidin (Cat. No. T7471). The microtubules were detected with a mouse monoclonal anti–ß-tubulin primary antibody and subsequently visualized with the green-fluorescent Alexa Fluor® 488 Goat Anti–Mouse IgG antibody (Cat. No. A11001). The image was contributed by Jonathan Zmuda, Immunomatrix, Inc.
Confocal micrograph of the cytoskeleton of a mixed population of granule neurons and glial cells. The F-actin was stained with red-fluorescent Texas Red®-X phalloidin (Cat. No. T7471). The microtubules were detected with a mouse monoclonal anti–ß-tubulin primary antibody and subsequently visualized with the green-fluorescent Alexa Fluor® 488 Goat Anti–Mouse IgG antibody (Cat. No. A11001). The image was contributed by Jonathan Zmuda, Immunomatrix, Inc.
Related Products
Catalog # Name Size Price (USD) Qty
A11001 Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488 1 mg
284.00
T7471 Texas Red™-X Phalloidin, Red, 591/608 nm 300 units
609.65
Online Exclusive
628.00
Save 18.35 (3%)
Related Images