Noyori Asymmetric Hydrogenation – an electrophilic addition reaction
In 1980, the Japanese Nobel prize-winning chemist Ryoji Noyori reported that BINAP (2,2'-bis(diphenylphosphino)-1,1'-binaphthyl) complexed with ruthenium [BINAP-Ru(II)] catalyzed the asymmetric hydrogenation of alpha-(acylamino)acrylic acids or esters to give the corresponding amino acid derivatives in excellent enantiomeric yields. Several years later, it was discovered that asymmetric hydrogenation of a wide variety of functionalized olefins could be achieved utilizing BINAP-Ru(II) dicarboxylate complexes. Subsequently, it was found that oligomeric halogen-containing BINAP-Ru(II) complexes were efficient catalysts for the asymmetric hydrogenation of functionalized ketones. Following these discoveries, the reduction of both functionalized olefins and ketones using BINAP-Ru(II) in the presence of hydrogen gas became known as the Noyori asymmetric hydrogenation.
Industrial uses of this technique include the synthesis of the anti-inflammatory drug naproxen and the antibacterial agent levofloxacin.
Mechanism of the Noyori asymmetric hydrogenation reaction
Review available Thermo Scientific products for the Noyori asymmetric hydrogenation reaction:
Catalog # Name Size Price (USD) Qty
B23785.MD (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98% Each
31.65
Online Exclusive
35.20
Save 3.55 (10%)
B23785.03 (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98% Each
74.65
Special offer
Online exclusive
Ends: 26-Sep-2025
87.80
Save 13.15 (15%)
B23785.06 (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98% Each
368.65
Special offer
Online exclusive
Ends: 30-Jun-2025
433.00
Save 64.35 (15%)
H26226.06 (+/-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 97+% Each
134.65
Online Exclusive
149.00
Save 14.35 (10%)
H26226.14 (+/-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 97+% Each
365.65
Special offer
Online exclusive
Ends: 26-Sep-2025
430.00
Save 64.35 (15%)
H26226.22 (+/-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 97+% Each
751.65
Special offer
Online exclusive
Ends: 30-Jun-2025
884.00
Save 132.35 (15%)
B23872.03 (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 97% Each
101.65
Special offer
Online exclusive
Ends: 26-Sep-2025
119.00
Save 17.35 (15%)
B23872.06 (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 97% Each
423.65
Online Exclusive
470.00
Save 46.35 (10%)
47182.09 -
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers.  Do you have a favorite reaction?  Let us know! 
Vote now