The Simmons-Smith Cyclopropanation is a prominent method of preparing cyclopropanes
Simmons-Smith Cyclopropanation Overview
In the Simmons-Smith cyclopropanation reaction, an olefin reacts with a zinc-copper couple and diiodomethane to form the corresponding cyclopropane.
Named Reactions in Organic Synthesis
Simmons-Smith Cyclopropanation Mechanism
Watch our video to learn more about the Simmons-Smith cyclopropanation and its mechanisms.
Video Player is loading.
Current Time 0:00
Duration 2:39
Loaded: 6.18%
Stream Type LIVE
Remaining Time 2:39
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Need reagents for the Simmons-Smith Cyclopropanation?
    Chem dex
    If you need further help to plan your synthesis, don’t miss our new app.  Chem dex is a new tool to explore hundreds of chemical reactions, and to have access to product recommendations. Visit: thermofisher.com/chemdex.
    Named Reactions eBook
    Named Reactions in Organic Synthesis eBook
    Download our Named Reactions eBook to learn more about the Simmons-Smith cyclopropanation. 
    Access 10 chapters of content about organic chemistry reactions that are named after the chemists that discovered them. This informative eBook includes quizzes so you can test your knowledge!
    History of the Simmons-Smith Cyclopropanation
    In 1958, Howard Ensign Simmons Jr. and Ronald D. Smith stereospecifically converted unfunctionalized alkenes such as cyclohexene to cyclopropanes using diiodomethane (CH2I2) in the presence of a zinc-copper couple. This reaction proved to be general, and has since become an important method of preparing cyclopropanes; it is known today as the Simmons-Smith cyclopropanation.
    The zinc-copper couple can be prepared by reacting zinc powder with copper sulfate solution. However, a zinc-silver couple often gives better yields and shorter reaction times. The use of diethylzinc with diiodomethane also gives excellent results, and is known as the Furukawa modification. The use of iodo- or chloromethyl samarium iodide for the cyclopropanation of allylic alcohols in the presence of other olefins is known as the Molander modification.
    The Simmons-Smith cyclopropanation reaction was used in the total synthesis of the antimitotic agent (+)-curacin A by the team of Shigeo Iwasaki to generate the cyclopropane ring.
    Other Electrophilic Addition Reactions
    Other Electrophilic Addition Reactions include: 
    For other types of reactions, visit our Named Reactions page.
    Acroseal Packaging
    Chemical reactions often involve the use of air- and moisture-sensitive solvents, and pyrophoric or hazardous reagents. Our AcroSeal packaging is a packaging solution designed to enable safe handling of these types of materials which are used in a variety of research and development applications, including NMR analysis and studies in drug discovery, agrochemicals, flavors and fragrances, and more. 
    Watch our video for more information. 
    Video Player is loading.
    Current Time 0:00
    Duration 4:00
    Loaded: 4.09%
    Stream Type LIVE
    Remaining Time 4:00