MF-FP-Compliance-Testing-470x250
When failure is not an option
Verify that your products can withstand electrostatic discharge (ESD) events and other threats with comprehensive simulators and test systems that ensure your semiconductors, integrated circuits, and other electronic products are in compliance with national, international, and industry ESD and electromagnetic compatibility (EMC) test standards.
Featured compliance testing and related products
The Thermo Scientific MK.1TE is a relay-based, exceptionally fast ESD and Static Latch-Up Test System used in the evaluation of advanced IC devices.  It fully addresses today’s JEDEC/ESDA standards, and can be configured with 64, 128, 192 or 256 test pins.
The Thermo Scientific MK.2TE is a relay-based, exceptionally fast ESD and Latch-Up Test System used in the evaluation of advanced IC devices.  It fully addresses today’s JEDEC/ESDA standards, and can be configured with 128, 256, 384, or 768 test pins.
The Thermo Scientific MK.4TE ESD & Latch-Up test system is a complete, robust and feature-filled turn-key instrumentation test package, which performs automatic and manual HBM, MM, and Latch-Up tests on devices with pin counts up to 2304 pins. It features the highest speed of test execution, lowest Zap interval, and extensive parallelism that enables concurrent Zapping with interleaved Trace test capability to global and company driven quality standards.
The Thermo Scientific Orion3 is a floor-standing, bench height robotic Charged Device Model (CDM) tester designed to meet all popular CDM test standards, allowing both Field Induced air discharge methods (FICDM) and contact (CCDM) methods like our patented CDM2 test method.
The Thermo Scientific Celestron is a two terminal bench-top TLP / VF-TLP test system for fast, accurate, reliable, and affordable characterization of advanced semiconductor structures.
The Thermo Scientific Pegasus 2-pin ESD Test System is a low parasitic tester platform used in the evaluation of advanced IC devices at both wafer and package levels. Determination of ESD failure thresholds is made easy using one of the available ESD waveforms and built-in curve tracing capabilities.
Thermo Scientific Test Fixture Boards are custom designed to your devices precise specifications.  Years of experience in constructing these boards ensures the highest quality waveforms.  Designs can be turned around quickly allowing minimal delays in beginning qualification.  All boards are guaranteed to operate correctly or we will repair or replace them free of charge.
Support
Contact us for service and calibration on Thermo Scientific and Keytek products. Call 1-978-275-0800 or 800-753-9835.