Magnetic Sector GC-HRMS Highlights
For over a decade, the global scientific community has chosen the DFS Magnetic Sector GC-HRMS for its superior performance in routine quantitative analysis of dioxins and POPs. Today, the DFS Magnetic Sector GC-HRMS continues to set the gold standard in providing robust performance and high-confidence results.
Robust sensitivity
Based on magnetic sector technology, the DFS Magnetic Sector GC-HRMS performs dioxin and POPs routine analysis, delivering ultimate confidence no matter the sample type or matrix. It features a large-volume ion source and high-capacity turbo pump for maximum sensitivity and robustness.
Global Compliance
Both food import-export control and environmental analyses require compliance with numerous government and analytical methods. The DFS Magnetic Sector GC-HRMS is the only GC-MS system compliant with existing official methods and regulations, including U.S. EPA, EU, and JIS methods.
Confidence
High-linearity detection and matrix-independent calibrations combined with automated mass calibration for each analysis run provide confidence in your analytical results. Additionally, the system displays the best dioxin installation specifications during each system installation (based on 2378-TCDD with 4 sigma noise definition) and makes LOD/LOQ calculations using S/N real-time detection limits calculated for each sample analysis.
Traceability
The DFS Magnetic Sector GC-HRMS features lock mass trace for constant monitoring of instrument stability and sample matrix effect in compliance with official methods, such as EPA 1613. Additionally, mass resolution is documented in a log file for each analysis, providing proof for stable selectivity.
Flexibility and productivity
The flexible configuration of the DFS Magnetic Sector GC-HRMS offers a unique dual GC configuration with the option to install up to 4 columns. The system can operate with different reference gases, e.g., PFK, FC43, etc.
Easy to use and maintain
The ion source can be exchanged without breaking vacuum for quick maintenance. Additionally, a separation valve enables venting of the ion source housing when necessary.
DFS Magnetic Sector GC-HRMS
Delivering confidence and robustness, the DFS Magnetic Sector GC-HRMS meets your current and future needs for worldwide compliance in dioxins and POPs analysis.
Learn more
Magnetic Sector GC-HRMS Videos
Maximized productivity for dioxin and PCB analysis
Curious to know 6 facts about Magnetic Sector Mass Spectrometers and Thermo Scientific DFS Magnetic Sector GC-HRMS for Dioxin analysis? Watch the series and let us know more facts!
A visit to NofaLab Laboratories
Meet with the team at NofaLab Laboratories (Rotterdam), where the team explains their dioxin analysis workflow with Magnetic Sector GC-HRMS and how important is fast turnaround time for their customers.
A visit to Pacific Rim Lab in Vancouver, Canada, with the DFS Magnetic Sector GC-HRMS product team
Follow the DFS Magnetic Sector GC-HRMS Product Team in their visit at Pacific Rim Lab, a very productive Dioxins and POPs lab.
Maximized productivity for dioxin and PCB analysis
Curious to know 6 facts about Magnetic Sector Mass Spectrometers and Thermo Scientific DFS Magnetic Sector GC-HRMS for Dioxin analysis? Watch the series and let us know more facts!
A visit to NofaLab Laboratories
Meet with the team at NofaLab Laboratories (Rotterdam), where the team explains their dioxin analysis workflow with Magnetic Sector GC-HRMS and how important is fast turnaround time for their customers.
A visit to Pacific Rim Lab in Vancouver, Canada, with the DFS Magnetic Sector GC-HRMS product team
Follow the DFS Magnetic Sector GC-HRMS Product Team in their visit at Pacific Rim Lab, a very productive Dioxins and POPs lab.
Resources
Related Products
Ordering Information
Contact us
* Required field
*
*
*
*
*
*
*
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific™ products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
Style Sheet for Global Design System
Style Sheet for Komodo Tabs