A close up image of the BioTitan retention device
Enhance the reliability and integrity of tubing assemblies
Experience the latest in retention technology for your single-use assemblies—the Thermo Scientific BioTitan Retention Device, an innovative replacement for traditional cable ties that enhances the reliability and integrity of assemblies with a 360-degree seal. With zero leaks reported since its 2021 launch, the BioTitan Retention Device offers superior performance. Take a 3D product tour to explore its benefits and features firsthand.
Contact us  Request sample  View 3D interactive tool
Key advantages and benefits
The BioTitan retention device replaces the need for cable ties
Legacy Cable Ties
BioTitan retention device on tubing
BioTitan Retention Device
The BioTitan retention device replaces the need for cable tiesUneven compression—Cable ties do not provide a 360⁰ seal around tubing and fittings and can also cause uneven compression, creating weak spots in the connection.BioTitan retention device on tubingSecure 360º compression system—Proprietary collar design and automated application process significantly reduces risk of fluid leaks and connection failures.
The BioTitan retention device replaces the need for cable tiesSharp edges—Cable ties are stiff and can have sharp edges after assembly. These edges and points can snag tubing assemblies, rip gloves, or potentially pierce BPC films adding risk to your process.BioTitan retention device on tubingNo sharp edges—Eliminates the need for secondary processes and minimizes risk of damage to BPCs.
The BioTitan retention device replaces the need for cable tiesPackaging— Extra bubble wrap is needed for packaging around the cable tie surface due to the sharp edges and points that could damage BPCs.BioTitan retention device on tubingSimplified user experience— Save time during setup with fewer unpacking steps and less packaging waste.
Watch to learn more about BioTitan Retention Device
Video Player is loading.
Current Time 0:00
Duration 2:03
Loaded: 0.00%
Stream Type LIVE
Remaining Time 2:03
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    BioTitan Retention Device Overview
    Video Player is loading.
    Current Time 0:00
    Duration 1:48
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 1:48
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      BioTitan Device Unpacking Video
      Video Player is loading.
      Current Time 0:00
      Duration 1:03
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 1:03
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        BioTitan Device Pressure Features
        The BioTitan Retention Device is featured on the following products:        
        Bioprocessing resources
        Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
        For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.