Labtainer Pro BPC with BioTitan Retention Device
Your bioprocessing workflow, reinvented
As technology and innovation advance within the bioprocessing industry, single-use technologies have also made considerable progress in the drug and vaccine manufacturing space. Some of the well-established and well-known advantages of single-use systems are lowered costs, reduced contamination risks, decreased facility footprint, increased flexibility, and production throughput efficiency with less cleanup; all resulting in quicker turnaround, and increased production capabilities. 
Key features of the Labtainer Pro BPC
Improving single-use product performanceBioProcess Container in-process inspection methods
  • Now manufactured with BioTitan Retention Device—designed to provide superior universal retention that minimizes the risk of leaks and failures at the connection point.
  • Proven film materials—available in both Aegis5-14 and CX5-14 films which are both produced in a cleanroom facility.
  • Enhanced port design and customization—configurable options available with two, three, or four ports without the need for excess ports on the chamber resulting in fewer potential failure points and no liquid holdup.
  • Improved sealing method—manufacturing process incorporates impulse heat sealing for port insertion utilizing Labtainer Automated Manufacturing (LAM).
  • Multiple in-process inspection methods
    • 100% automated manufacturing inspection
    • 100% visual inspection
    • 100% integrity test on all standard configurations
Striving to provide the highest standard of quality and deliver Upgraded Labtainer Pro BPC outer packaging
the best product in the industry
  • 100% integrity testing—helium integrity is conducted on all standard configurations.
  • Lot-based testBacterial endotoxin testing (BET) is an in-vitro assay for detection and quantitation of each test to the USP <85> standard. Particulate analysis testing (PAT) includes procedures for removing, counting, and sizing particulate contaminants on or in the BPC to the USP <788> standard.
  • Improved outer polyethylene (PE) bag—the protective PE bags used to package the Labtainer Pro BPC are manufacturing in a controlled environment, resulting in cleaner packaging with less risk of contaminants and particulate accumulation.
  • Upgraded packaging—the outer packaging of the Labtainer Pro BPC has been further developed to prevent damage due to unpacking with sharps or other tools, as well as reduce outer packaging material by approximately 25%.
Designed to optimize the overall user experienceLabtainer Pro BPC improved handle feature
  • Enhanced ergonomics—the improved handle feature allows for more ergonomic manipulation. An additional lower handle is available on the 2, 5, 10, and 20 L units.
  • Improved drainage—the shape of the Labtainer Pro BPC has been redesigned to allow for better drainage with less manipulation required, featuring chamber geometry that tapers to the ports. This reduces the holdup column at the bottom of the BPC.
  • User-friendly packaging—featuring easy-peel tape and easy-tear outer PE bags with directional-tear removal, which maintains the product integrity and provides a particulate barrier prior to product removal.
Improving single-use product performanceBioProcess Container in-process inspection methods
  • Now manufactured with BioTitan Retention Device—designed to provide superior universal retention that minimizes the risk of leaks and failures at the connection point.
  • Proven film materials—available in both Aegis5-14 and CX5-14 films which are both produced in a cleanroom facility.
  • Enhanced port design and customization—configurable options available with two, three, or four ports without the need for excess ports on the chamber resulting in fewer potential failure points and no liquid holdup.
  • Improved sealing method—manufacturing process incorporates impulse heat sealing for port insertion utilizing Labtainer Automated Manufacturing (LAM).
  • Multiple in-process inspection methods
    • 100% automated manufacturing inspection
    • 100% visual inspection
    • 100% integrity test on all standard configurations
Striving to provide the highest standard of quality and deliver Upgraded Labtainer Pro BPC outer packaging
the best product in the industry
  • 100% integrity testing—helium integrity is conducted on all standard configurations.
  • Lot-based testBacterial endotoxin testing (BET) is an in-vitro assay for detection and quantitation of each test to the USP <85> standard. Particulate analysis testing (PAT) includes procedures for removing, counting, and sizing particulate contaminants on or in the BPC to the USP <788> standard.
  • Improved outer polyethylene (PE) bag—the protective PE bags used to package the Labtainer Pro BPC are manufacturing in a controlled environment, resulting in cleaner packaging with less risk of contaminants and particulate accumulation.
  • Upgraded packaging—the outer packaging of the Labtainer Pro BPC has been further developed to prevent damage due to unpacking with sharps or other tools, as well as reduce outer packaging material by approximately 25%.
Designed to optimize the overall user experienceLabtainer Pro BPC improved handle feature
  • Enhanced ergonomics—the improved handle feature allows for more ergonomic manipulation. An additional lower handle is available on the 2, 5, 10, and 20 L units.
  • Improved drainage—the shape of the Labtainer Pro BPC has been redesigned to allow for better drainage with less manipulation required, featuring chamber geometry that tapers to the ports. This reduces the holdup column at the bottom of the BPC.
  • User-friendly packaging—featuring easy-peel tape and easy-tear outer PE bags with directional-tear removal, which maintains the product integrity and provides a particulate barrier prior to product removal.
BioTitan retention device
BioTitan device offers 360° of universal retention 
This industry-leading product is also now manufactured with BioTitan Retention Device—for heightened performance and lower risk. The BioTitan device is designed to provide superior universal retention that minimizes the risk of leaks and failures at the connection point. The innovative design concept of the Thermo Scientific Labtainer Pro BioProcess Container (BPC) provides flexibility and assurance without compromise
Request information   Download data sheet
Automated manufacturing
BioProcess Container automated manufacturing 
Combining technology and innovation
We create products driven by industry expertise and a history of developing best-in-class solutions. To support an increasing demand for robust, high-quality products, and prompted by the development of the Labtainer Pro BPC, we created Labtainer Automated Manufacturing (LAM) technology. Standard and customized Labtainer Pro BPCs are manufactured using LAM technology.
LAM technology elevates Labtainer Pro BPC manufacturing to help ensure a product that is unsurpassed in terms of quality, reliability, and ease of use. The benefits of automated manufacturing include increased product consistency, a decreased likelihood of manufacturing errors, a safer manufacturing environment, and optimized production process efficiencies.          
Available accessories
Labtainer Pro BPC Accessories 
The Labtainer Pro BPC has been engineered with multiple product accessories to help maximize productivity and to help improve the overall experience of working with the product
  • The Labtainer Pro tote provides BPC protection and access to lines without the need to open the lid. The tray inside the tote is designed with a tilted plate to allow for optimal drainage. This product is constructed of medical-grade, high-density PE.
Labtainer Pro BPC Interactive
Using this interactive tool, browse the key features and benefits of the Labtainer Pro BPC. Interact with the fill and dispense feature while seeing a 360° view of all available sizes ranging from 50 mL to 20 L.
Labtainer Pro BPC Interactive Tour
Ordering information
Catalog # Name Size Price (USD) Qty
PL30021.11 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 2 ports, end-ported with plugs 2 L -
PL30022.11 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 2 ports, end-ported with body luer lock and MPC insert 2 L -
PL30023.11 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 3 ports, end-ported with MPC body, MPC insert, and body needleless luer lock 2 L -
PL30024.11 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 3 ports, end-ported with AseptiQuik G connectors and body needleless luer lock 2 L -
PL30025.11 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 3 ports, end-ported with AseptiQuik G connector, body luer lock, and body needleless luer lock 2 L -
PL30026.11 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 4 ports, end-ported with MPX, tri-clamps, and body needleless luer lock 2 L -
PL30008.11 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 2 ports, end-ported with plugs 2 L -
PL30009.11 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 2 ports, end-ported with body luer lock and MPC insert 2 L -
PL30010.11 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 3 ports, end-ported with MPC body, MPC insert, and body needleless luer lock 2 L -
PL30011.11 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 3 ports, end-ported with AseptiQuik G connectors and body needleless luer lock 2 L -
PL30012.11 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 3 ports, end-ported with AseptiQuik G connector, body luer lock, and body needleless luer lock 2 L -
PL30013.11 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 4 ports, end-ported with MPX, tri-clamps, and body needleless luer lock 2 L -
Catalog # Name Size Price (USD) Qty
PL30021.12 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 2 ports, end-ported with plugs 5 L -
PL30022.12 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 2 ports, end-ported with body luer lock and MPC insert 5 L -
PL30023.12 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 3 ports, end-ported with MPC body, MPC insert, and body needleless luer lock 5 L -
PL30024.12 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 3 ports, end-ported with AseptiQuik G connectors and body needleless luer lock 5 L -
PL30025.12 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 3 ports, end-ported with AseptiQuik G connector, body luer lock, and body needleless luer lock 5 L -
PL30026.12 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 4 ports, end-ported with MPX, tri-clamps, and body needleless luer lock 5 L -
PL30008.12 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 2 ports, end-ported with plugs 5 L -
PL30009.12 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 2 ports, end-ported with body luer lock and MPC insert 5 L -
PL30010.12 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 3 ports, end-ported with MPC body, MPC insert, and body needleless luer lock 5 L -
PL30011.12 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 3 ports, end-ported with AseptiQuik G connectors and body needleless luer lock 5 L -
PL30012.12 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 3 ports, end-ported with AseptiQuik G connector, body luer lock, and body needleless luer lock 5 L -
PL30013.12 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 4 ports, end-ported with MPX, tri-clamps, and body needleless luer lock 5 L -
Catalog # Name Size Price (USD) Qty
PL30021.13 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 2 ports, end-ported with plugs 10 L -
PL30022.13 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 2 ports, end-ported with body luer lock and MPC insert 10 L -
PL30023.13 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 3 ports, end-ported with MPC body, MPC insert, and body needleless luer lock 10 L -
PL30024.13 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 3 ports, end-ported with AseptiQuik G connectors and body needleless luer lock 10 L -
PL30025.13 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 3 ports, end-ported with AseptiQuik G connector, body luer lock, and body needleless luer lock 10 L -
PL30026.13 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 4 ports, end-ported with MPX, tri-clamps, and body needleless luer lock 10 L -
PL30008.13 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 2 ports, end-ported with plugs 10 L -
PL30009.13 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 2 ports, end-ported with body luer lock and MPC insert 10 L -
PL30010.13 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 3 ports, end-ported with MPC body, MPC insert, and body needleless luer lock 10 L -
PL30011.13 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 3 ports, end-ported with AseptiQuik G connectors and body needleless luer lock 10 L -
PL30012.13 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 3 ports, end-ported with AseptiQuik G connector, body luer lock, and body needleless luer lock 10 L -
PL30013.13 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 4 ports, end-ported with MPX, tri-clamps, and body needleless luer lock 10 L -
Catalog # Name Size Price (USD) Qty
PL30008.15 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 2 ports, end-ported with plugs 15 L -
PL30010.15 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 3 ports, end-ported with MPC body, MPC insert, and body needleless luer lock 15 L -
PL30011.15 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 3 ports, end-ported with AseptiQuik G connectors and body needleless luer lock 15 L -
PL30012.15 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 3 ports, end-ported with AseptiQuik G connector, body luer lock, and body needleless luer lock 15 L -
PL30013.15 Labtainer™ Pro BioProcess Container with BioTitan Retention, CX5-14, 4 ports, end-ported with MPX, tri-clamps, and body needleless luer lock 15 L -
PL30021.15 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 2 ports, end-ported with plugs 15 L -
PL30022.15 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 2 ports, end-ported with body luer lock and MPC insert 15 L -
PL30023.15 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 3 ports, end-ported with MPC body, MPC insert, and body needleless luer lock 15 L -
PL30024.15 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 3 ports, end-ported with AseptiQuik G connectors and body needleless luer lock 15 L -
PL30025.15 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 3 ports, end-ported with AseptiQuik G connector, body luer lock, and body needleless luer lock 15 L -
PL30026.15 Labtainer™ Pro BioProcess Container with BioTitan Retention, Aegis™ 5-14, 4 ports, end-ported with MPX, tri-clamps, and body needleless luer lock 15 L -
Catalog # Name Size Price (USD) Qty
PL30021.04 Labtainer™ Pro BioProcess Container, Aegis™ 5-14, 2 ports (plugs) 20 L -
PL30008.04 Labtainer™ Pro BioProcess Container, CX5-14, 2 ports (plugs) 20 L -
PL30022.04 Labtainer™ Pro BioProcess Container, Aegis™ 5-14, 2 ports (body luer lock and MPC insert) 20 L -
PL30009.04 Labtainer™ Pro BioProcess Container, CX5-14, 2 ports (body luer lock and MPC insert) 20 L -
PL30023.04 Labtainer™ Pro BioProcess Container, Aegis™ 5-14, 3 ports (MPC body, MPC insert, and body needleless luer lock) 20 L -
PL30010.04 Labtainer™ Pro BioProcess Container, CX5-14, 3 ports (MPC body, MPC insert, and body needleless luer lock) 20 L -
PL30024.04 Labtainer™ Pro BioProcess Container, Aegis™ 5-14, 3 ports (AseptiQuik G connectors and body needleless luer lock) 20 L -
PL30011.04 Labtainer™ Pro BioProcess Container, CX5-14, 3 ports (AseptiQuik G connectors and body needleless luer lock) 20 L -
PL30025.04 Labtainer™ Pro BioProcess Container, Aegis™ 5-14, 3 ports (AseptiQuik G connector, body luer lock, and body needleless luer lock) 20 L -
PL30012.04 Labtainer™ Pro BioProcess Container, CX5-14, 3 ports (AseptiQuik G connector, body luer lock, and body needleless luer lock) 20 L -
PL30026.04 Labtainer™ Pro BioProcess Container, Aegis™ 5-14, 4 ports (MPX, tri-clamps, and body, needleless luer lock) 20 L -
PL30013.04 Labtainer™ Pro BioProcess Container, CX5-14, 4 ports (MPX, tri-clamps, and body needleless luer lock) 20 L -
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
For Research Use or Further Manufacturing.  Not for diagnostic use or direct administration into humans or animals.