Scanning electron microscopy (SEM) provides high-quality surface and cross-sectional imaging of 3D structures. As semiconductor feature sizes continue to decrease in response to consumer and industry demands, nanoscale metrology becomes an increasingly important contributor to process and device design yield. Accurate and repeatable nanoscale measurements of critical dimensions require high-resolution SEM imaging and extremely accurate magnification calibration.
The ultra-high-resolution automated SEM capabilities offered by Thermo Fisher Scientific, when combined with our industry-leading software solutions, perform imaging and metrology with the automation, precision, and robustness needed to meet the challenges of leading memory and logic customers. Leveraging NIST-traceable calibration standards, mature imaging automation (both cross-sectional and top-down), and next-generation machine-learning-enabled metrology, Thermo Scientific automated SEMs provide cost-effective 3D metrology for direct process and device monitoring, as well as faster time to market.
Thermo Fisher Scientific offers a range of SEM metrology tools for critical dimension analysis, including the Thermo Scientific Verios 5 SEM with Thermo Scientific AutoSEM Software for top-down SEM metrology. Please click through to the appropriate product pages below for more information, or to request a demo.
SEM metrology performed automatically with Thermo Scientific software.
SEM metrology performed automatically with Thermo Scientific software.
SEM metrology workflow examples
Applications
Semiconductor-RD_1920x1080
Semiconductor research and development
Innovation starts with research and development. Learn more about solutions to help you understand innovative structures and materials at the atomic level.
Semi-Materials-Character_1920x1080
Semiconductor materials characterization
Many factors impact yield, performance, and reliability. Learn more about solutions to characterize physical, structural, and chemical properties.
Semi-Metrology_1920x1080
Semiconductor metrology
Manufacturing today’s complex semiconductors requires exact process controls. Learn more about advanced metrology and analysis solutions to accelerate yield learnings.
Semiconductor Failure Analysis
Semiconductor Failure Analysis
Complex semiconductor device structures result in more places for defects to hide. Learn more about failure analysis solutions to isolate, analyze, and repair defects.
Semiconductor power devices
Semiconductor power devices
Novel architectures and materials pose new challenges. Learn how to pinpoint faults and characterize materials, structures, and interfaces.
Display Module Failure Analysis
Semiconductor display technology
Display technologies are evolving to improve display quality and light conversion efficiency. Learn how metrology, failure analysis, and characterization solutions provide insights.
Samples
Semiconductor Materials and Device Characterization
As semiconductor devices shrink and become more complex, new designs and structures are needed. High-productivity 3D analysis workflows can shorten device development time, maximize yield, and ensure that devices meet the future needs of the industry.
Learn more ›
Style Sheet to change H2 style to p with em-h2-header class
Products
Style Sheet for Instrument Cards Original
Helios 5 HX/Helios 5 UX/Helios 5 FX DualBeam
  • Fully automated, high-quality, ultra-thin TEM sample preparation
  • High throughput, high resolution subsurface and 3D characterization
  • Rapid nanoprototyping capabilities
Thermo Scientific Verios 5 XHR scanning electron microscope (SEM)
Verios 5 XHR SEM
  • Monochromated SEM for sub-nanometer resolution over the full 1 keV to 30 keV energy range
  • Easy access to beam landing energies as low as 20 eV
  • Excellent stability with piezo stage as standard
Contact us
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
semiconductors
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Learn more ›