Organism TypeMouseHAS-, a mouse mammary carcinoma mutant cell line, which is defective in hyaluronan synthase activity. Polyoma large T antigen-expressing cell lines were established by transfecting HAS mutant cells with pdl3027 plasmid containing polyoma T antigen gene as described previously(14) . Polyoma large T antigen-mediated replication of plasmids in these cell lines was assessed by measurement of the methylation status of the plasmid DNA (15) . Finally, a clonal cell line named HAS-P was chosen for a transient expression cloning.
TissuesBreast/Mammary
PhenotypeAdherent
Primaryyes
ApplicationStable Cell Transfection
Products
Catalog # Name Size Price (USD) Qty
18292011 Lipofectin™ Transfection Reagent, 1 mL Each
710.00
Every step of the way, a wide range of cell health products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.