Human Corneal Epithelial cells treated with Chloroquine to induce autophagy
Gibco® Human Corneal Epithelial cells were treated with 30uM Chloroquine overnight. The following day the cells were fixed and permeabilized and labeled with 0.5ug/mL anti-LC3B with a goat anti mouse Alexa Fluor® 488 secondary (Green) Cells were counter stained with 1ug/mL Hoechst 33342 (Blue) Images were acquired with a Molecular Devices ImageXpress High content imager
Gibco® Human Corneal Epithelial cells were treated with 30uM Chloroquine overnight. The following day the cells were fixed and permeabilized and labeled with 0.5ug/mL anti-LC3B with a goat anti mouse Alexa Fluor® 488 secondary (Green) Cells were counter stained with 1ug/mL Hoechst 33342 (Blue) Images were acquired with a Molecular Devices ImageXpress High content imager
Related Products
Catalog # Name Size Price (USD) Qty
L10382 LC3B Antibody Kit for Autophagy Each
555.65
Online Exclusive
574.00
Save 18.35 (3%)
A11034 Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488 1 mg
342.00
Related Images