Organelle Labeling in Live Cells
Human osteosarcoma (U-2 OS) cells were labeled with CellMask™ Deep Red plasma membrane stain (Cat. no. C10046), the endoplasmic reticulum-selective ER-Tracker™ Green dye (Cat. no. E34251), mitochondrion-selective MitoTracker® Red CMXRos dye (Cat. no. M7512), and blue-fluorescent NucBlue™ Live Cell Stain (Cat. no. R37605). Labeled cells were imaged in the Live Cell Imaging Solution (Cat. no. A124291DJ) on the Zeiss 710 point-scanning spectral detection confocal microscope.
Human osteosarcoma (U-2 OS) cells were labeled with CellMask™ Deep Red plasma membrane stain (Cat. no. C10046), the endoplasmic reticulum-selective ER-Tracker™ Green dye (Cat. no. E34251), mitochondrion-selective MitoTracker® Red CMXRos dye (Cat. no. M7512), and blue-fluorescent NucBlue™ Live Cell Stain (Cat. no. R37605). Labeled cells were imaged in the Live Cell Imaging Solution (Cat. no. A124291DJ) on the Zeiss 710 point-scanning spectral detection confocal microscope.
Related Products
Catalog # Name Size Price (USD) Qty
C10046 CellMask™ Deep Red Plasma Membrane Stain, 100 μL Each
332.65
Online Exclusive
343.00
Save 10.35 (3%)
E34251 ER-Tracker™ Green (BODIPY™ FL Glibenclamide), for live-cell imaging Each
516.00
M7512 MitoTracker™ Red CMXRos - Special Packaging, 20 x 50 μg, 20 x 50 μg Each
506.65
Online Exclusive
522.00
Save 15.35 (3%)
R37605 NucBlue™ Live ReadyProbes™ Reagent (Hoechst 33342) Each
179.65
Online Exclusive
195.00
Save 15.35 (8%)
Related Images